Sigma-Aldrich
Analytical / Chromatography > Hot Topics > July 2009
hot topics banner
Hot Topics in Analytical & Chromatography - July 2009
In this issue, we address the following topics:
  • Antiobesity Drugs
  • Pharmaceutical Products in Water
  • Multi-Residue Pesticide Analysis
back to top
Antiobesity DrugsAntiobesity Drugs - The health implications of the near-epidemic obesity problem have made research aimed at treating obesity a hot topic.
Sibutramine, one of a few drugs approved for long-term treatment of obesity, is a chiral compound. Because the (R) and (S) enantiomers have opposing effects, it is a perfect example of the importance of chiral HPLC to pharmaceutical and clinical analysts.
HPLC Separation of Sibutramine Enantiomers
Both Astec CHIROBIOTIC V and V2 successfully separate sibutramine enantiomers, using a simple, MS-friendly mobile phase. The larger α value on the V2 would make it more suitable for preparative applications.
Sibutramine Separation
Conditions
Columns: Astec CHIROBIOTIC™ V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
Astec CHIROBIOTIC™ V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
Mobile phase: 15 mM ammonium formate in methanol
Flow rate: 1 mL/min.
Det.: UV at 230 nm
Temp.: 25°C
Sample: Sibutramine in methanol (1 mg/mL), Inj. vol.: 5 µL
For more information, visit the CHIROBIOTIC web page.
back to top
Pharmaceutical Products in WaterPharmaceutical Products in Water - There has been increasing concern about the long-term consequences of improperly disposed pharmaceuticals reaching the public’s drinking water supply.
Supel-Select HLB SPE is a new hydrophilic polymer SPE amenable for use with NEW EPA Method 1694: Pharmaceuticals & Personal Care Products in Water, Soil, Sediments, and Biosolids
Supel-Select HLB SPE is a hydrophilic modified polymer SPE phase developed for the extraction of a wide array of compounds from aqueous samples.
Features and Benefits
  • Priced lower than the major competitors
  • Amenable to generic methodology
  • Reduce ion-suppression
  • Resistant to over-drying, increasing reproducibility
  • Low UV & MS extractables
  • Stringent production & QC guidelines
  • Greater capacity for smaller elution volumes
Supel-Select HLB
Additional Information:
Visit sigma-aldrich.com/spe
View Supelco Reporter 27.3 (1.3mb pdf)
back to top
Multi-Residue Pesticide AnalysisMulti-Residue Pesticide Analysis - The surveillance of pesticide residues in agricultural and dairy products has been a major concern for government agencies and industrial organizations worldwide.
Dispersive SPE, also known as QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe), has become the preferred sample prep technique for pesticide analysis.
Supelco's Dispersive SPE comprises of a line of centrifuge tubes packed with a pre-determined amounts of salts and SPE sorbents to support the most common methods use today.
Features and Benefits
  • Configured for CEN Standard Method EN 15662 & AOAC Method 2007.01
  • Pre-packed for convenience and time savings
  • Centrifuge tubes are leak-free
  • Packed with the highest quality salts, buffering agents, and SPE phases
  • Comprehensive customization services available
Dispersive SPE Tubes
Additional Information:
Visit sigma-aldrich.com/quechers