Analytical / Chromatography > Hot Topics > September 2009
hot topics banner
Hot Topics in Analytical & Chromatography - September 2009
In this issue, we address the following topics:
    back to top
Vapor IntrusionVapor Intrusion - Old problem, new threat: Ground level contamination from underground storage tanks (UST) and industrial run-off is a fast-growing residential and commercial building health issue that has made Vapor Intrusion a Hot Topic.
Supelco offers the most comprehensive range of carbon-based adsorbents (Carbopack and Carbotrap) and carbon molecular sieves (Carboxen) to help analysts meet their difficult vapor intrusion sampling needs.
Vapor Intrusion monitoring requires different types of sampling media to investigate contamination by diverse compounds, including chlorinated VOCs like trichloroethylene (TCE) and vinyl chloride (VC), as well as BTEX, PAHs and PCBs. Supelco offers a wide range of both active and passive sampling media collection devices with our adsorbent technology inside to collect and measure these compounds.
Additional Information:
  1. Visit www.sigma-aldrich.com/vapor-intrusion
  2. Learn more about Supelco's range of adsorbents for air monitoring
    back to top
On-demand web seminarsOn-demand Web Seminars - Interested in learning more about HPLC? Don’t have time to attend training sessions? Supelco has seminars available on the web, available 24 hours a day, 7 days a week for your viewing.
HPLC Web seminars cover topics such as HPLC Theory, column fundamentals, use of alternate selectivity, and high speed separations. Helpful videos demonstrating the use of our sample prep and Karl Fischer titration products are also available on demand.
Additional Information:
  1. Visit www.sigma-aldrich.com/videos
  2. If you prefer to attend regional seminars in person, check out our regional seminar schedule at www.sigma-aldrich.com/analytical-seminars
    back to top
Fast GC Fast GC - In today’s analytical laboratory, it is becoming increasingly desirable to increase sample throughput without sacrificing quality.
The principles of Fast GC can be applied to do exactly this.
Fast GC was successfully used to reduce the analysis time for the GC-MS analysis of 86 semivolatile analytes in just 8.2 minutes. This analysis typically requires 15-20 minutes using conventional GC.
Fast GC Separation
Peak IDs
   1. n-Nitrosodimethylamine 30. Naphthalene-d8 (I.S.) 59. 2,4,6-Tribromophenol (surr.)
2. Pyridine 31. Naphthalene 60. 4-Bromophenyl phenyl ether
3. 2-Fluorophenol (surr.) 32. 4-Chloroaniline 61. Hexachlorobenzene
4. Phenol-d6 (surr.) 33. Hexachlorobutadiene 62. Pentachlorophenol
5. Phenol 34. 4-Chloro-3-methylphenol 63. Phenanthrene-d10 (I.S.)
6. Aniline 35. 2-Methylnaphthalene 64. Phenanthrene
7. Bis(2-chloroethyl)ether 36. Hexachlorocyclopentadiene 65. Anthracene
8. 2-Chlorophenol-d4 (surr.) 37. 2,4,6-Trichlorophenol 66. Carbazole
9. 2-Chlorophenol 38. 2,4,5-Trichlorophenol 67. Di-n-butyl phthalate
10. 1,3-Dichlorobenzene 39. 2-Fluorobiphenyl (surr.) 68. Fluoranthene
11. 1,4-Dichlorobenzene-d4 (I.S.) 40. 2-Chloronaphthalene 69. Benzidine
12. 1,4-Dichlorobenzene 41. 2-Nitroaniline 70. Pyrene
13. Benzyl alcohol 42. Dimethyl phthalate 71. Terphenyl-d14 (surr.)
14. 1,2-Dichlorobenzene-d4 (surr.) 43. 2,6-Dinitrotoluene 72. 3,3’-Dimethylbenzidine
15. 1,2-Dichlorobenzene 44. Acenaphthylene 73. Butylbenzyl phthalate
16. 2-Methylphenol 45. 3-Nitroaniline 74. 3,3’-Dichlorobenzidine
17. Bis(2-chloroisopropyl)ether 46. Acenaphthene-d10 (I.S.) 75. Benzo(a)anthracene
18. n-Nitroso-di-n-propylamine 47. Acenaphthene 76. Bis(2-ethylhexyl)phthalate
19. 4-Methylphenol 48. 2,4-Dinitrophenol 77. Chrysene-d12 (I.S.)
20. Hexachloroethane 49. 4-Nitrophenol 78. Chrysene
21. Nitrobenzene-d5 (surr.) 50. Dibenzofuran 79. Di-n-octyl phthalate
22. Nitrobenzene 51. 2,4-Dinitrotoluene 80. Benzo(b)fluoranthene
23. Isophorone 52. Diethyl phthalate 81. Benzo(k)fluoranthene
24. 2-Nitrophenol 53. 4-Chlorophenyl phenyl ether 82. Benzo(a)pyrene
25. 2,4-Dimethylphenol 54. Fluorene 83. Perylene-d12 (I.S.)
26. Bis(2-chloroethoxy)methane 55. 4-Nitroaniline 84. Indeno(1,2,3-cd)pyrene
27. Benzoic acid 56. 2-Methyl-4,6-dinitrophenol 85. Dibenzo(a,h)anthracene
28. 2,4-Dichlorophenol 57. n-Nitrosodiphenylamine 86. Benzo(g,h,i)perylene
29. 1,2,4-Trichlorobenzene 58. Azobenzene    
Conditions
Columns: SLB-5ms, 20 m x 0.18 mm I.D., 0.18 µm (28564-U)
Oven: 40 °C (0.7 min.), 55 °C/min. to 240 °C, 28 °C/min. to 330 °C (2 min.)
Inj.: 250 °C
MSD interface.: 330 °C
Scan range: m/z 40-450
Carrier gas: helium, 40 cm/sec, constant
Injection: 0.5 µL, 10:1 split
Liner: 2 mm I.D., fast FocusLiner™ inlet liner with taper (2879501-U)
Sample: 80 component semivolatile standard at 50 ppm plus 6 internal standards (at 40 ppm) in methylene chloride
Additional Information:
Practical considerations, theoretical discussions, a listing of columns in Fast GC dimensions, multiple chromatograms, plus a list of literature for additional reading can be found in the Fast GC Brochure (2.5Mb PDF).