Sigma-Aldrich
Analytical / Chromatography > HPLC > Accessories > JETSTREAM PLUS
HPLC Accessories
JETSTREAM PLUS
Astec Products | Technical Resources
Featujetstreamres
  • Full management of temperature
  • Enhanced separations
  • Improves reproducibility
  • Increases performance of analysis
  • Reliable results
  • Increased chiral resolution
JETSTREAM PLUS offers full management of temperature enhanced separations, improving the reproducibility and increasing the performance of your analysis. In addition, JETSTREAM PLUS, a multiple Peltier column thermostat with heating/cooling capability, has two-way forced air circulation. The 5-85°C and 0-80°C temperature range is controlled by new dual reference sensor technology and the auto-calibration feature guarantees reliable results. The setting of fixed temperatures, steps and gradients is done with a simple numerical keypad and a two line character display.
The JETSTREAM PLUS Peltier heating/cooling HPLC column thermostat is fully programmable for isothermal, stepwise and linear ramp calculation. Temperature can be input in Celsius or Fahrenheit. The compact unit holds up to 5 columns in lengths up to 40 cm. It includes an integrated leak detector, adjustable sensitivity and acoustic control column protection by temperature lock feature. An RS-232 computer link is included as part of the control circuitry.
Specifications
Temp. Range: 5-85 °C or 0-85 °C
Temp. Accuracy: ± 0.2 °C
Temp. Stability: ± 0.1 °C
Power: 100-245 VAC 50/60 Hz 100 W
Dimensions: 135(W) x 310(L) x 450(H) mm
Weight: 8 Kg
Temperature Effects on Resolution
jetstream_temp_eff1 jetstream_temp_eff2
For ordering information, go to our On-Line Catalog