HPLC Accessories
Mobile Phase Degassing & Filters
Are you tired of waiting for a noisy baseline to settle following a column change? To help ensure a steady baseline and consistent analytical results, Supelco® offers a complete line of mobile phase filtration and degassing products.
Supelco Mobile Phase Degassing System
Supelco Mobile Phase Filtration Apparatus 1
Supelco Mobile Phase Filtration Apparatus 2
Mobile Phase Reservoir System
Inlet Filters
Supelco Mobile Phase Degassing System
  • 4 Channel
  • 0-5 ml/min Flow Range
  • Validation output satisfies system compliance
  • Smart sensor detects leaks and communicates with vacuum pump
  • LED indicates degassing status
  • Unprecedented 4 year warranty
The Supelco Mobile Phase Degassing system with its smart sensor not only detects and alerts you to leaks, it also communicates with the vacuum pump. If a change in vacuum is detected due to mobile phase flow rate changes, the pump can compensate by changing its speed. The validation output records vacuum level to satisfy system compliance and validation requirements. The degassing system has a Teflon® AF membrane, with NO-OX™ fittings and tubing.
Supelco Mobile Phase Filtration Apparatus 1
Connects to 1000 mL sidearm flask (not included)
Includes:
  • 250 mL glass reservoir
  • Funnel base and stopper
  • Clamp
  • Stainless steel holder and screen
  • 10 Teflon gaskets
  • 50 Nylon 66 filters (47 mm, 0.45 µm pores)
Supelco Mobile Phase Filtration Apparatus 2
Connects to 1000 mL sidearm flask (not included)
Includes:
  • 250 mL glass reservoir
  • TS 34/45 tapered funnel base
  • TS 34/45 tapered 1000 mL flask and glass cap
  • Clamp
  • Stainless steel holder and screen
  • 10 Teflon gaskets
  • 50 Nylon 66 filters (47 mm, 0.45 µm pores)
(Flask not included)
Mobile Phase Reservoir System
This convenient system is made entirely of inert parts: glass, Tefzel, PEEK, and TFE Teflon. Includes 10 ft./3 m of 1/8 in. O.D. Teflon tubing, a PEEK bottom-of-the-bottle filter/sparger, a plug, and a 1-liter or 2-liter safety-coated bottle.
10 µm Slip-On Inlet Filter
We recommend this filter for solvent inlet applications. The 10 µm filter protects the HPLC system from contamination while minimizing pump cavitation problems. Compatible with the 1.5 mm I.D. tubing used with many HPLC pumps. Also fits the 2.2 mm I.D. tubing used with Varian pumps and the 3.0 mm I.D. tubing used with Waters pumps. 316 stainless steel tip and filter element connected by a KEL-F collar.
Description Product No.
10 µm Slip-On Inlet Filter 59277
2 µm Stainless Steel Inlet Filter
All stainless steel construction with a 2 µm porous filter element, 11/16 in. / 1.7 cm diameter.
1/8 in. pipe.
Description Product No.
2 µm Stainless Steel Inlet Filter 58267