Sigma-Aldrich
Analytical / Chromatography > HPLC > Columns > Ascentis Express
HPLC
Ascentis Express
image for fused core hplc column
Ascentis Express columns provide a breakthrough in HPLC column performance.
HPLC Fused-Core Webinar
Watch a presentation
on Fused-Core HPLC
Based on Fused-Core™ particle technology, Ascentis Express provides the high speed and high efficiency of sub-2-µm particles, but at half of the backpressure. This lower pressure means that Ascentis Express can be run on all HPLC systems, including conventional as well as UHPLC systems.
KEY USER BENEFITS
There are five key user benefits with the Ascentis Express HPLC Columns:
1. Hyper-Fast Separations
  • Designed for high flow rates and sample throughput, especially LC-MS
  • Half of the backpressure of sub-2-µm particles
2. High-Definition Resolution
  • Twice the efficiency of 3 µm particles
  • Longer columns permit doubling the plates over sub-2-µm particles
  • Shortest analyte diffusion path
3. Super-Sensitive
  • Sharper peaks provide better Signal:Noise
4. Super-Rugged
  • Narrowest particle size distribution
  • 2µm porosity frits like most rugged conventional columns
5. A Family of selectivities that rivals the competition
  • C18, RP-Amide, Phenyl-Hexyl, C8, HILIC
  • Match competitor selectivitiy and performance at lower pressure
Available in C18, C8, RP-Amide, HILIC, (NEW!) Phenyl-Hexyl; also nano/capillary sizes
Ascentis Express C18 and C8 provide classic reversed-phase selectivity with performance benefits for Fast HPLC and High Resolution HPLC. Ascentis Express HILIC provides all of the benefits of Fused-Core particle technology to the emerging technique of HILIC mode chromatography. Ascentis Express RP-Amide and Phenyl-Hexyl provides alternate selectivity to reversed-phase separations.
Ordering information for Ascentis Express HPLC columns
Technical Resources
Sign up to receive the Fused-Core Report! A quarterly report on the lastest applications, publications, and new products related to fused-core particle technology.
Publication Title Type Size (kb) No. Pages
Ascentis Express Brochure (view / request) Brochure 1.1Mb 8
NEW! Ascentis Express HPLC Resource Guide Brochure 2.1Mb 28
NEW! Ascentis Express Phenyl-Hexyl HPLC Columns Product Info. 714 2
Ascentis Express RP-Amide HPLC Columns Product Info. 536 2
Ascentis Express HILIC HPLC Columns Flyer 360 2
Ascentis Express Capillary HPLC Columns with Fused-Core Technology Product Info. 320 2
Save Acetonitrile with Ascentis Express HPLC Columns Product Info. 636 2
Approaches to Lessening the Impact of the Acetonitrile Shortage on Your Reversed-Phase HPLC Separations Tech Report 197 3
High Performance HPLC Fittings/Interconnects Product Info. 304 2
NEW! Frit Porosity: An Important Factor to Consider Before Using Sub-2µm UHPLC Columns Tech Report 671 2
Improving HPLC Performance: Relationship between particle size, column efficiency and column pressure App Note 183 556 3
Ascentis Express: A Fused-Core™ Particle HPLC Column for High Speed and High Efficiency Separations with Low Back Pressures App Note 181 91 2
Guide to Dispersion Measurement Data Sheet 60 1
Guidelines for Optimizing Performance with Ascentis Express HPLC Columns Data Sheet 31 1
Improving USP methods using Ascentis Express (Reporter volume 26.1) Article 482 3
Fast HPLC using Ascentis Express (Reporter volume 25.5) Article 181 3
High Resolution HPLC Using Ascentis Express (TheReporter volume 25.4) Article 307 2
Transferring Methods Using Ascentis Express (TheReporter volume 25.3) Article 273 2
Introducing Ascentis Express (TheReporter volume 25.2) Article 482 3
Trademarks
Ascentis - Sigma-Aldrich Co.
Fused-Core - Advanced Materials Technology, Inc.
UPLC - Waters Corp.