Ascentis® Express 5 µm HPLC Columns
A new choice for improving performance of traditional HPLC systems!
Ascentis Express columns, based on 5 µm Fused-Core® particles, offer the benefits of faster and more efficient separations compared to columns based on 3 µm and 5 µm totally porous particles, without any backpressure concerns. Ascentis Express 5 µm columns are also extremely rugged, making them an ideal choice for bioanalytical LC-MS methods since they excel under the demands of high flow rate and high throughput.
On-demand webinar:
Introducing Ascentis Express 5 µm
HPLC Columns
Ordering Information
Request an evaluation column
(Eligibility criteria apply)
Ascentis Express
5 µm Brochure (view)
Available Phases: C18, F5, C8, Phenyl-Hexyl, ES-Cyano , HILIC, OH5, and RP-Amide
High Efficiency Particles
5 µm Ascentis Express Fused-Core particles provide double the number of plates per pressure relative to 5 µm totally porous particles, and four times the number of plates per pressure relative to 3 µm totally porous particles.
Best Plates per Pressure
Comparison of Particle Characteristics
Particle Type Shell
Thickness (µm)
BET Surface
Area (m2/g)
Average Pore
Diameter (Å)
Plates* Pressure*
(bar)
Plates per
Pressure
5 µm Ascentis Express Fused-Core 0.6 90 90 28,300 78 363
3 µm totally porous N/A 300 100 24,200 309 78
5 µm totally porous A N/A 300 100 14,600 100 146
5 µm totally porous B N/A 170 120 14,400 63 229
5 µm totally porous C N/A 450 100 15,300 120 128
* Plates and Pressure: reported for 1-chloro-4-nitrobenzene at the flow rate corresponding to the plate height minimum
Kinetic Plots - Comparison of Particles
The 5 µm Fused-Core particle outperforms 3 µm and 5 µm totally porous particles under all conditions.
As a result, Ascentis Express 5 µm columns outperform popular 3 µm and 5 µm columns (based on totally porous particles), which are currently used in most HPLC systems.
Better Performance - Faster & More Efficient Separation
The chromatograms below show a relative comparison of the separation obtained using an Ascentis Express 5 µm column vs that obtained using a popular column based on 5 µm totally porous particles; identical method conditions were used.
Ascentis Express C18 5 µm, 15 cm x 4.6 mm I.D.
N=23,422 for last peak, 80 Bar, run time = 8 min
Conditions
Mobile Phase:  50:50 water:acetonitrile
Flow Rate:  1 mL/min
Detection:  UV, 250 nm
Sample:  Mix of polar and non-polar analytes
Analytes:  1. Uracil
2. Acetophenone
3. Benzene
4. Toluene
5. Naphthalene
Popular C18 5 µm, 15 cm x 4.6 mm I.D.
N=13,633 for last peak, 72 Bar, run time = 16 min
Trademarks:
Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.