|
Ascentis Express Fused-Core® HPLC Columns
Fused-Core Technology
|
Ascentis Express HPLC columns, based on Fused-Core particle technology, provide more than twice the speed and efficiency of traditional columns at half the backpressure of sub-2-µm columns. This performance enhancement is applicable to all HPLC instruments (in addition to UHPLC systems).
Why do columns based on Fused-Core technology offer superior performance? The Fused-Core particle has three characteristic features over the traditional porous particle, resulting in a number of performance benefits - especially for Fast HPLC and UHPLC applications.
Features of Fused-Core particles over traditional porous particles
- Narrower particle size distribution
- More consistent packed bed
- Shorter diffusion path
Performance Benefits
- More than double the speed of current methods
- Increased resolution over current methods
- Super rugged columns compared to sub-2-µm
- Added sensitivity (from sharper peaks)
- Easily transferable methods, from UHPLC to HPLC
For an in-depth explanation, with practical examples that demonstrate the performance benefits of Fused-Core technology, watch the presentation linked on this page.
|
 (Ascentis Express home page)
|
| Comparison of particle size distribution, packed bed consistency, and diffusion path length for Fused-Core particles vs. traditional porous silica particles |
| Fused-Core Particles |
|
Traditional Porous Particles |
 |
|
 |
The innovative manufacturing process for Fused-Core particles produces a very narrow particle size distribution. A narrow particle size distribution allows for the use of large porosity frits that resist clogging, resulting in a more rugged column. Traditional porous particles are not manufactured in a way to yield extremely narrow particle size distributions. |
 |
|
 |
| The “A” term in the van Deemter equation accounts for the effects of inhomogeneities in the packed bed of an HPLC column. Narrow particle size distributions form a more consistent packed bed and a consistent path length, minimizing analyte diffusion through the column. This eddy diffusion is effectively independent of mobile phase velocity. |
 |
|
 |
| The short diffusion path of the Fused-Core particle yields sharper peaks than traditional porous particle columns. The minimized resistance to mass transfer, the “C” term in the van Deemter equation, of the Fused-Core particle provides sharper peaks than traditional porous particles. The short diffusion path also permits the use of higher flow rates without peak broadening. |
|
Trademarks Fused-Core - Advanced Materials Technology, Inc.
|