Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > HPLC > Columns > Chiral > CHIROBIOTIC V2
HPLC Columns
CHIROBIOTIC V2
Astec Products | Technical Resources | Chiral Services
Ordering Information
Features
  • New extension for the CHIROBIOTIC series
  • Enhanced selectivity and capacity in the polar organic mode and polar ionic mode
  • Capacity increases of 2 to 20 fold over standard CHIROBIOTIC phases
  • Most beneficial for basic molecules
  • Neutral molecules can often be run in pure methanol
The polar organic and the polar ionic modes have become increasingly popular largely because of their ideal compatibility with MS detection and preparative LC. Two years of extensive research at Astec in understanding these mechanisms has resulted in new possibilities for extending their effectiveness even further. Important changes in linkage chemistries and the silica and the silica have now led to the new CHIROBIOTIC V2 and T2 chiral stationary phases – columns of choice for those really difficult separations, and especially, for preparative LC.
These new phases extend the capabilities of the series. They are designed to be used in the optimization process since they offer some distinct advantages in the polar ionic mode and polar organic mode. The standard CHIROBIOTIC phases remain the columns of choice for method development screening and for reversed phase separations.