Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > HPLC > Columns > Chiral > Astec CYCLOBOND
Chiral HPLC Columns
Astec CYCLOBOND
Astec Products | Technical Resources | Bibliography | Chiral Services
New! icon Frequently Asked Questions - over 60 topics to assist you with chiral chromatography!
CYCLODEXTRIN Stationary Phases for Chiral Separations and Highly Selective Achiral Separations
CYCLOBOND is the name given to Astec technology for bonding cyclodextrins to a high purity silica gel through a stable ether linkage. Introduced in 1983, this patented stationary phase retains ability to form inclusion complexes, and allows for numerous chemical separations by selectively including into the cyclodextrin cavity, from solution, a wide variety of organic molecules. CYCLOBOND I are β-cyclodextrin bonded and CYCLOBOND II are γ-cyclodextrin bonded.
CYCLOBOND I 2000 series of HPLC columns is specially formulated to meet today’s stringent requirements for analysis in the pharmaceutical industry, and for small analytes of general interest in the chemical and environmental areas. Astec has focused on the need to accurately and reproducibly separate enantiomers. The result is a high performance range of chiral separation phases with guaranteed batch to batch reproducibility, greater stability and improved selectivity and resolution. Based on the original CYCLOBOND I (β-cyclodextrin) columns, the CYCLOBOND I 2000 series are second-generation products. The native β-cyclodextrin and eight β-cyclodextrin derivatives are in the CYCLOBOND I 2000 series.
CYCLOBOND II series columns are excellent chiral selectors for multi-ring structures such as those based on anthracene, chrysene or pyrene. These are γ-cyclodextrin bonded phases, and consist of 8 glucopyranose units arranged in the same truncated cone shape. Applications include steroids, porphorins, FMOC amino acids. Applications for CYCLOBOND II AC, an acetylated derivative, include napthylethylamine, steroids, and sterols.
Phases (ordering information)
CYCLOBOND I 2000
CYCLOBOND I 2000 AC
CYCLOBOND I 2000 SP
CYCLOBOND I 2000 RSP
CYCLOBOND I 2000 HP RSP   
CYCLOBOND I 2000 DMP
CYCLOBOND I 2000 DM
CYCLOBOND I 2000 DNP
CYCLOBOND II
CYCLOBOND II AC
CYCLOBOND Method Development Kit