Sigma-Aldrich
Analytical / Chromatography > HPLC > Columns > Discovery® > BIO Wide Pore C18
HPLC
BIO Wide Pore C18
Discovery BIO logo
Ordering Information
Discovery BIO Wide Pore C18
HPLC Columns and Capillaries
Bonded Phase:   Covalently-bonded octadecylsilane (C18), endcapped
Silica:   Spherical, 3µm, 5µm and 10µm diameters, 300Å pore diameter,
    high purity (<10ppm metals)
Column IDs:   0.18mm to 21.2mm
Solutions for Protein and Peptide Separation Challenges
•
Proteomics and biotherapeutics: high resolution of synthetic peptides and peptide maps
•
LC/MS; no-bleed, capillary dimensions, and pH stability
•
Peptide purification: Scalable from analytical to preparative
•
Reproducible separations batch-to-batch and column-to-column
Here's the
Proof
MATCHED SELECTIVITY FROM ANALYTICAL TO PREP
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 3µm; 15cm x 4.6mm, 5µm; 15cm x 10mm, 10µm, Mobile Phase: (A) 80:20, (0.1% TFA in H2O) : (0.1%TFA in MeCN), (B) 66:34, (0.1% TFA in H2O) : (0.1%TFA in MeCN), Flow Rate: 6.02cm/min, Temp: 30°C, Detection: 215nm, Sample: Peptide Mix (Sigma Cat. No. P2693)
Column Parameters & Run Conditions:
Column
CV(mL) Injection(µL) Flow(mL/min)
15cm x 4.6mm, 3µm
1.64 5.0 1.00
15cm x 4.6mm, 5µm
1.71 5.2 1.00
15cm x 10mm, 10µm
8.01 24.5 4.73
Gradient:
CV %A %B
0 100 0
2 100 0
9 0 100
g001511_13
PEPTIDE MAP OF
CARBOXYMETHYLATED APOHEMOGLOBIN TRYPTIC DIGEST
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 5µm; Mobile Phase: (A) 95:5, (0.1% TFA in H2O):(0.1% TFA in MeCN); (B) 50:50, (0.1% TFA in H2O):(0.1% TFA in MeCN); Flow Rate: 1mL/min; Temp: 30°C; Detection: 215nm; Injection: 50µL; Sample: caboxymethylated apohemoglobin tryptic digest; Gradient: 0-100% B in 65 min
g001510
STABILITY OF DISCOVERY BIO WIDE PORE C18
AT ALKALINE pH
Alkaline pH Stability
Retention times of the sample components remained constant after 29,000 bed volumes were passed through the column.
Column: Discovery BIO Wide Pore C18, 5cm x 4.6mm, 5µm; Mobile Phase: 25mM potassium phosphate, pH 8 : MeOH (95:5); Flow Rate: 2mL/min; Temp: 35°C; Detection: 254nm
g001520
UNDETECTABLE LC/MS BLEED
LC/MS Bleed Analysis on Discovery BIO Wide Pore C18
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 3µm; Mobile Phase: (A) 0.1% TFA in H2O, (B) 0.1% TFA in MeOH; Flow Rate: 1mL/min; Temp: 30°C; Gradient: 0-100% B in 15 min, hold for 5 min, then back to 0% B for 10 min
No TIC baseline rise relative to blank run (gradient run w/o column)
g001522
g001523
LOT TO LOT REPRODUCIBILITY OF DISCOVERY BIO WIDE PORE C18
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 5µm; Mobile Phase: 80% 10mM ammonium acetate, pH 6.8 : 20% MeOH; Flow Rate: 1mL/min; Temp: 35°C; Detection: 254nm; Injection: 5µL
1. Uracil (8µg/mL)
2. Procainamide (30µg/mL)
3. Sorbic acid (4µg/mL)
4. Pyridine (100µg/mL)
5. Caffeine (40µg/mL)
6. Phenol (240µg/mL)
g001514_6
Ordering Information
3µm Discovery BIO Wide Pore C18 HPLC Columns
ID(mm) Length(cm) Cat. No.
0.18 5 65603-U
0.18 10 65604-U
0.32 5 65526-U
0.32 10 65527-U
0.5 5 65517-U
0.5 10 65518-U
1.0 5 65504-U
1.0 10 65506-U
2.1 5 567200-U
2.1 10 567201-U
2.1 15 567202-U
4.6 5 567203-U
4.6 10 567204-U
4.6 15 567205-U
5µm Discovery BIO Wide Pore C18 HPLC Columns
ID(mm) Length(cm) Cat. No.
0.18 5 65606-U
0.18 10 65607-U
0.18 15 65608-U
0.32 15 65529-U
0.5 15 65519-U
1.0 15 65508-U
1.0 25 65509-U
2.1 5 568200-U
2.1 10 568201-U
2.1 15 568202-U
2.1 25 568203-U
4.0 5 568210-U
4.0 10 568211-U
4.0 15 568212-U
4.0 25 568213-U
4.6 5 568220-U
4.6 10 568221-U
4.6 15 568222-U
4.6 25 568223-U
10.0 25 568230-U
10µm Discovery BIO Wide Pore C18 HPLC Columns
ID(mm) Length(cm) Cat. No.
4.6 25 567206-U
10.0 5 567207-U
10.0 15 567208-U
10.0 25 567209-U
21.2 5 567210-U
21.2 15 567211-U
21.2 25 567212-U
2cm Supelguard Cartridges with Discovery BIO C18 Packings
  Description 2.1mm ID 4.0mm ID
  3µm Particle Size
  2 Pack
567270-U 567272-U
  Kit triangle
567271-U 567273-U
  5µm Particle Size
  2 Pack
568270-U 568272-U
  Kit triangle
568271-U 568273-U
  For 4.0mm ID or 4.6mm ID analytical columns
triangle Kit includes a holder and one replaceable cartridge.
1cm Supelguard Cartridges with Discovery BIO C18 Packings
  Description 10mm ID
  10µm Particle Size 567282-U