Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > HPLC > Columns > Discovery® > BIO Wide Pore C8
HPLC
BIO Wide Pore C8
Discovery BIO logo
Ordering Information
Discovery BIO Wide Pore C8
HPLC Columns and Capillaries
Bonded Phase:   Covalently-bonded octylsilane (C8), endcapped
Silica:   Spherical, 3µm, 5µm and 10µm diameters, 300Å pore diameter,
    high purity (<10ppm metals)
Column IDs:   0.32mm to 21.2mm
Solutions for Protein and Peptide Separation Challenges
Proteomics and biotherapeutics: high resolution of synthetic peptides and peptide maps
LC/MS; no-bleed, capillary dimensions, and pH stability
Peptide purification: Scalable from analytical to preparative
Intermediate hydrophobicity makes it a good method development starting point
Here's the
Proof
RESOLUTION OF ANGIOTENSINS AT NEUTRAL pH
Human Angiotensins I, II, & III contain a seven amino acid sequence conserved between the three types. Differences between the three peptides vary only by three amino acid residues. Angiotensin II differs from Angiotensin III by an N-terminal aspartate; and Angiotensin I contains an N-terminal asparate as well as two C-terminal amino acids (histidine and leucine). At low pH conditions (most typical for peptide chromatography), angiotensins II & III are not resolved since the asparate residue contains no charge (pH 2.0). Therefore, resolution of the two angiotensins is typically conducted at alkaline pH. Note however that baseline resolution was achieved at pH 7 using Discovery BIO Wide Pore C8.
Column:
1. Angiotensin II (1.67g/L)
2. Angiotensin III (1.67g/L)
3. Angiotensin I (1.67g/L)
D R V Y I H P F    
  R V Y I H P F    
D R V Y I H P F H L
Discovery BIO Wide Pore C8, 15cm x 4.6mm, 5µm; Mobile Phase: (A) 10mM NH4H2PO4/ NH4OH, pH 7; (B) 50:50, (20mM NH4H2PO4/ NH4OH, pH 7):MeCN; Flow Rate: 1mL/min; Temp: 30°C; Detection: 215nm; Injection: 6µL; Gradient: 30-60% B in 15 min
g001503
INTERMEDIATE HYDROPHOBICITY
OF DISCOVERY BIO WIDE PORE C8
Because of its intermediate hydrophobicity (less hydrophobic than C18; more hydrophobic than C5), Discovery BIO Wide Pore C8 is an ideal phase for early method development.
Column: Discovery BIO Wide Pore C18, C8, & C5, 15cm x 4.6mm, 5µm; Mobile Phase: (A) 95:5, (0.1% TFA in H2O):(0.1% TFA in MeCN); (B) 50:50, (0.1% TFA in H2O):(0.1% TFA in MeCN); Flow Rate: 1mL/min; Temp: 30°C; Detection: 215nm; Injection: 50µL; Sample: caboxymethylated apohemoglobin tryptic digest; Gradient: 0-100% B in 65 min
g001504_06
STABILITY OF DISCOVERY BIO WIDE PORE C8
AT ALKALINE AND ACIDIC pH
Alkaline pH Stability
Retention times of the sample components remained constant after 21,000 bed volumes were passed through the column.
Column: Discovery BIO Wide Pore C8, 5cm x 4.6mm, 5µm; Mobile Phase: 25mM potassium phosphate, pH 8 : MeOH (95:5); Flow Rate: 2mL/min; Temp: 35°C; Detection: 254nm
g001508
Acidic pH Stability
Retention times of the sample components remained constant after 40,000 bed volumes were passed through the column.
Column: Discovery BIO Wide Pore C8, 5cm x 4.6mm, 5µm; Mobile Phase: (A) 0.1% TFA in 5% MeCN; (B) 0.1% TFA in 75% MeCN; Flow Rate: 2mL/min; Temp: 35°C; Detection: 220nm; Gradient: 0-22% B in 22min
g001509
RESOLUTION OF INSULINS FROM VARIOUS SPECIES
Insulin is composed of two peptide chains: a 21 amino acid A chain and a B chain of 30 amino acids. The two chains are linked via two disulfide bonds, and much of the sequence is conserved across species (including the locations of the disulfide bonds) allowing for a very similar three dimensional conformation regardless of the species type. For example, porcine insulin differs from human by only one amino acid, and bovine differs by three amino acids. In the application, Discovery BIO Wide Pore C8 was able to resolve all three types of insulin.
Column:
1. Bovine insulin (5µg)
2. Human insulin (5µg)
3. Porcine insulin (5µg)
Discovery BIO Wide Pore C8, 15cm x 4.6mm, 5µm; Mobile Phase: (A) 71:29, (0.1% TFA in H2O):(0.1% TFA in MeCN); (B) 68:32, (0.1% TFA in H2O):(0.1% TFA in MeCN); Flow Rate: 1mL/min; Temp: 30°C; Detection: 215nm; Injection: 50µL; Gradient: 0-100% B in 30min
g001507
Ordering Information
3µm Discovery BIO Wide Pore C8 HPLC Columns
ID(mm) Length(cm) Cat. No.
NEW 0.32, 0.50, 1.0
  inquire
2.1 5 567213-U
2.1 10 567214-U
2.1 15 567215-U
4.6 5 567216-U
4.6 10 567217-U
4.6 15 567218-U
5µm Discovery BIO Wide Pore C8 HPLC Columns
ID(mm) Length(cm) Cat. No.
NEW 0.32, 0.50, 1.0
  inquire
2.1 5 568300-U
2.1 10 568301-U
2.1 15 568302-U
2.1 25 568303-U
4.0 5 568310-U
4.0 10 568311-U
4.0 15 568312-U
4.0 25 568313-U
4.6 5 568320-U
4.6 10 568321-U
4.6 15 568322-U
4.6 25 568323-U
10.0 25 568330-U
10µm Discovery BIO Wide Pore C8 HPLC Columns
ID(mm) Length(cm) Cat. No.
4.6 25 567219-U
10.0 5 567220-U
10.0 15 567221-U
10.0 25 567222-U
21.2 5 567223-U
21.2 15 567224-U
21.2 25 567225-U
2cm Supelguard Cartridges with Discovery BIO C8 Packings
  Description 2.1mm ID 4.0mm ID
  3µm Particle Size
  2 Pack
567274-U 567276-U
  Kit triangle
567275-U 567277-U
  5µm Particle Size
  2 Pack
568370-U 568372-U
  Kit triangle
568371-U 568373-U
For 4.0mm ID or 4.6mm ID analytical columns
triangle Kit includes cartridge, stand-alone holder, tubing, 2 nuts and ferrules.
1cm Supelguard Cartridges with Discovery BIO C8 Packings
  Description 10mm ID
  10µm Particle Size 567284-U