Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
HPLC
Cyano
Discovery logo
Ordering Information
Discovery Cyano HPLC Columns
Unique retention and selectivity enables better, faster separations
• Excellent peak shape and efficiency
• Unique selectivity compared to a C18
• Significantly less retentive than C18, reducing the amount of organic needed in the mobile phase
  Properties      
Bonded Phase:   Cyanopropylsilane, endcapped Cyano structure
Silica:   Spherical, high purity (Fe <20; NA <7; Ca <7;
Ti <1; AI <1; Mg <1ppm)
Particle Size:   5µm
Pore Size:   180(Å)
Surface Area:   200m2/g
%C:   4.5%
Coverage:   3-4µmoles/m2
2003cat_pg26a
2003cat_pg26b
Ordering Information
5µm Discovery Cyano HPLC Columns
ID(mm) Length(cm) Cat. No.
NEW! 2.1             2
(call to order) 577513-U
NEW! 2.1             3
(call to order) 577514-U
2.1   5 59355-U21
2.1 10 569521-U
2.1     12.5 569524-U
2.1 15 59356-U21
NEW! 3.0             2
(call to order) 577515-U
NEW! 3.0             3
(call to order) 577516-U
3.0   5 59355-U30
3.0 10 569522-U
3.0    12.5 569525-U
3.0 15 59356-U30
3.0 25 59357-U30
4.0   5 59355-U40
4.0 10 569523-U
4.0    12.5 569526-U
4.0 15 59356-U40
4.0 25 59357-U40
NEW! 4.6             2
(call to order) 577517-U
NEW! 4.6             3
(call to order) 577518-U
4.6   5 59355-U
4.6 10 569520-U
4.6    12.5 569527-U
4.6 15 59356-U
4.6 25 59357-U
5µm Discovery Cyano Validation Packs*
ID(mm) Length(cm) Cat. No.
2.1 5 55715-U21
2.1 10 569806-U
2.1 15 55717-U21
4.6 5 55715-U
4.6 10 569807-U
4.6 15 55717-U
4.6 25 55719-U
* Packs include 3 columns, each from a different lof of bonded phase.
2cm Supelguard Cartridges with Discovery Cyano Packings
  Description 2.1mm ID 3.0mm ID 4.0mm ID
  5µm Particle Size
  2 Pack
59584-U 569571-U 59586-U
  Kit triangle
59583-U 569570-U 59585-U
For 4.0mm ID or 4.6mm ID analytical columns
triangle Kit includes cartridge, stand-alone holder, tubing, 2 nuts and ferrules.
How to Contact Supelco
In USA:
Phone: 800-247-6628
  or 814-359-3441
fax: 800-325-5052
email: techservice@sial.com
Outside the USA: click here