|
| |
| Discovery Cyano HPLC Columns |
| |
| Unique retention and selectivity enables better, faster separations |
• Excellent peak shape and efficiency • Unique selectivity compared to a C18 • Significantly less retentive than C18, reducing the amount of organic needed in the mobile phase |
| |
| Properties |
|
|
|
| Bonded Phase: |
|
Cyanopropylsilane, endcapped |
 |
| Silica: |
|
Spherical, high purity (Fe <20; NA <7; Ca <7; Ti <1; AI <1; Mg <1ppm) |
| Particle Size: |
|
5µm |
| Pore Size: |
|
180(Å) |
| Surface Area: |
|
200m2/g |
| %C: |
|
4.5% |
| Coverage: |
|
3-4µmoles/m2 |
|
| |
| |
 |
 |
| Ordering Information |
| 5µm Discovery Cyano HPLC Columns |
|
|
| 5µm Discovery Cyano Validation Packs* |
|
|
| 2cm Supelguard Cartridges with Discovery Cyano Packings |
| |
Description |
2.1mm ID |
3.0mm ID |
4.0mm ID  |
| |
5µm Particle Size |
| |
|
59584-U |
569571-U |
59586-U |
| |
| |
Kit  |
|
59583-U |
569570-U |
59585-U |
| |
 |
For 4.0mm ID or 4.6mm ID analytical columns |
 |
Kit includes cartridge, stand-alone holder, tubing, 2 nuts and ferrules. |