|
| |
| LC-MS Columns |
| |
| Discovery® HPLC Columns for Fast LC-MS Analysis |
| New: HS F5 available in short column format. |
| |
| The Discovery LC-MS line offers short length columns for fast LC-MS analysis that are rugged and dependable. Supelco offers a robust product that withstands column shock due to fast gradient turnaround times. Columns installed on LC-MS instruments are typically operated using gradients of 5%-95% organic with gradient cycle times of 2-3 minutes. Our column lifetime is maximized through the use of stable bonding chemistry, high purity silica and optimal proprietary packing procedures. |
| |
The Discovery line of HPLC columns offers excellent retentivity, selectivity and reproducibility for optimal results. LC-MS formats are now available with Discovery C18, C8, Cyano and HS F5 phases.
Read the applications report "Sensitive LC-Analysis of Basic Compounds Using Discovery HS F5" found in TheReporter Vol. 22.5. |
| |
| Discovery LC-MS Columns Deliver... |
|
|
|
| Properties |
|
C18 |
|
C8 |
| Bonded Phase: |
|
Octadecylsilane, endcapped |
|
Octylsilane, endcapped |
| Silica: |
|
Spherical, high purity, <10ppm metals |
|
Spherical, high purity, <10ppm metals |
| Particle Size: |
|
5µm |
|
5µm |
| Pore Size: |
|
180Å |
|
180Å |
| Surface Area: |
|
200m2/g |
|
200m2/g |
| %C: |
|
12% |
|
7.5% |
| Coverage: |
|
3µmoles/m2 |
|
3.4µmoles/m2 |
| |
 |
| |
| Properties |
|
CYANO |
|
HS F5 |
| Bonded Phase: |
|
Cyanopropylsilane, endcapped |
|
Pentafluorophenylpropyl, endcapped |
| Silica: |
|
Spherical, high purity, <10ppm metals |
|
Spherical, high purity, <10ppm metals |
| Particle Size: |
|
5µm |
|
3.5µm |
| Pore Size: |
|
180Å |
|
120Å |
| Surface Area: |
|
200m2/g |
|
300m2/g |
| %C: |
|
4.5% |
|
12% |
| Coverage: |
|
3.5µmoles/m2 |
|
4µmoles/m2 |
| |
|
| High Speed Separations |
| |
| Discovery LC-MS columns can shorten analysis times to less than 4 minutes for the study of antibiotics. |
| Figure 1. Rifampicin on Discovery C18 |
| |
| |
|
|
| Column 1: |
|
Discovery C18, 15cm x 2.1mm ID, 5µm particles |
| Cat. No.: |
|
50495521 |
| Column 2: |
|
Discovery C18, 2cm x 2.1mm ID, 5µm particles |
| Cat. No.: |
|
577507-U |
| Mobile Phase: |
|
60:40, 33mM HCO2H:CH3CN |
| Flow Rate: |
|
0.2mL/min |
| Temp.: |
|
35ºC |
| Det.: |
|
ESI(+), full scan |
| Inj.1: |
|
6µL |
| Inj.2: |
|
0.8µL |
| Sample: |
|
10mg/L in 50% CH3OH |
|
| |
|
| |
| Figure 2. Application of Short, Fast LC-MS Columns for Analysis of Veterinary Antibiotics on Discovery C18 and C8 Columns |
| |
| |
|
|
| Column 1: |
|
Discovery C18, 2cm x 2.1mm ID, 5µm particles |
| Cat. No.: |
|
577507-U |
| Column 2: |
|
Discovery C8, 2cm x 2.1mm ID, 5µm particles |
| Cat. No.: |
|
577501-U |
| Mobile Phase: |
|
A=50mM HOAc/NH4OH, (pH 6.0), B=water, C=CH3OH |
| Flow Rate: |
|
0.2mL/min |
| Temp.: |
|
65ºC |
| Inj.: |
|
1µL |
| Det.: |
|
ESI(-) |
| Sample: |
|
|
| 1. |
|
Lasalocid A (1µg/mL) |
| 2. |
3. |
|
Salinomycin (10µg/mL) |
| 4. |
|
Narasin (5µg/mL) |
|
| |
| Discovery C18, A:B:C = 20:5:75 |
Discovery C8, A:B:C = 20:10:70 |
| |
|
| Excellent Peak Shape |
| |
| Figure 3. Application of Short, Fast LC-MS Columns for Analysis of Erythromycin c and Clarithromycin on Discovery C18, C8, and Cyano Columns |
| |
| |
|
|
| Column 1: |
|
Discovery C18, 2cm x 2.1mm ID, 5µm particles |
| Cat. No.: |
|
577507-U |
| Column 2: |
|
Discovery C8, 2cm x 2.1mm ID, 5µm particles |
| Cat. No.: |
|
577501-U |
| Column 3: |
|
Discovery Cyano, 3cm x 2.1mm ID, 5µm particles |
| Cat. No.: |
|
577514-U |
| Mobile Phase: |
|
A=50mM HCO2H, B=water, C=90:10 CH3OH:CH3CN |
| Flow Rate: |
|
0.2mL/min |
| Temp.: |
|
35ºC |
| Inj.: |
|
1µL |
| Det.: |
|
ESI(+) |
| Sample: |
|
|
| 1. |
|
Erythromycin c (1µg/mL) |
| 2. |
|
Clarithromycin (1µg/mL) |
|
| |
Discovery C18 A:B:C = 20:35:45 |
Discovery C8 A:B:C = 20:35:45 |
Discovery Cyano A:B:C = 20:40:40 |
| |
|
| Mechanical Stability via Fast Gradient Testing |
| |
| The Supelco Discovery LC-MS HPLC columns are rigorously tested in order to provide the highest mechanical stability under repeated gradient cycles. Table 1 shows of the impact on column efficiency, peak shape, retention times and back pressure after 1000 cycles of a 2-minute rapid (ballistic) gradient with 1-minute equilibration period. |
| |
| There is no loss of column efficiency or increase in tailing or column backpressure, and no change in retention time. See Table 2 for details of testing. |
| |
| Table 1. Rapid Gradient Turnaround Testing of Discovery LC-MS HPLC Columns |
| Phase |
Efficiency (N/column) |
USP Tailing Factor |
Retention Time (min.) |
Pressure (bar) |
| Discovery C18 (3cm x 3.0mm ID) |
| Before stability test |
2792 |
1.04 |
1.989 |
39 |
| After stability test |
2899 |
1.09 |
1.985 |
41 |
| |
| Discovery C8 (2cm x 2.1mm ID) |
| Before stability test |
1451 |
1.05 |
1.767 |
26 |
| After stability test |
1507 |
1.09 |
1.761 |
25 |
| |
| Discovery Cyano (3cm x 4.6mm ID) |
| Before stability test |
3329 |
1.01 |
1.368 |
44 |
| After stability test |
3448 |
1.01 |
1.362 |
40 |
|
| Table 2. Test Conditions |
| Discovery C18 (3cm x 3.0mm ID) |
| Flow: |
|
0.8mL/min |
| Gradient: |
|
10-90% methanol over 2 minutes back to 10% for 1 minute equilibrium for each 3 minute cycle |
| Temp.: |
|
30°C |
| Wavelength: |
|
254nm |
| Pressure: |
|
76bar for first cycle 75bar after last cycle |
| No. of Cycles: |
|
1,135 three-minute |
| Total Volume: |
|
2724mL |
| Void Volume: |
|
0.447 |
| |
| Discovery C8 (2cm x 2.1mm ID) |
| Flow: |
|
0.5mL/min |
| Gradient: |
|
10-90% methanol over 2 minutes back to 10% for 1 minute equilibrium for each 3 minute cycle |
| Temp.: |
|
30°C |
| Wavelength: |
|
254nm |
| Pressure: |
|
40bar after first cycle 38bar after last cycle |
| No. of Cycles: |
|
1,089 three-minute |
| Total Volume: |
|
1634mL |
| Void Volume: |
|
0.393 |
| |
| Discovery Cyano (3cm x 4.6mm ID) |
| Flow: |
|
0.2mL/min |
| Gradient: |
|
10-50% acetnonitrile over 2 minutes back to 10% for 1 minute equilibrium for each 3 minute cycle |
| Temp.: |
|
30°C |
| Wavelength: |
|
254nm |
| Pressure: |
|
72bar after first cycle 70bar after last cycle |
| No. of Cycles: |
|
937 three-minute |
| Total Volume: |
|
5622mL |
| Void Volume: |
|
0.434 |
|
|
|
| Discovery C18 Bleed Analysis |
| |
| Mass spectral responses from background ions often inhibit both qualitative and quantitative analysis in LC-MS experiments. Common sources of background ions include the solvents, interface or system contamination, and HPLC column bleed. The presence of background ions can be reduced by using high quality solvents such as LC-MS CHROMASOLV from Riedel-de Haën, maintaining a clean LC-MS system, and by using high quality HPLC stationary phases from Supelco. |
| |
| Supelco utilizes technologically advanced bonding chemistry and highly purified silica to minimize the potential for column bleed. |
| |
| LC-MS Bleed Analysis |
| The Total Ion Chromatogram (TIC) shown in the top of Figure 4 shows the result from an MS detector of one gradient cycle with a Discovery C18, 2cm x 2.0mm ID column in-line. The two selective ion chromatograms (SIC) in the bottom of Figure 4 show the mass spectrum taken from the shaded region of the TIC (at the high organic content point in the gradient). The bottom right SIC shows that the Discovery C18 column does not introduce any additional masses that were not in the blank run (bottom left SIC). The Discovery C18 column exhibits low bleed for LC-MS analyses. |
| |
| Total Ion Chromatogram |
| |
| Discovery C18, 2cm x 2.0mm ID, acetonitrile gradient in presence of 0.1% formic acid / 5 cycles / ESI (+) mode acquisition |
|
| Figure 4. LC-MS Bleed Study |
 |
| |
Blank Run (no LC column) (19-21 min.) |
Discovery C18 column (19-21 min.) |
 |
Product Profile - Discovery LC-MS Columns - (T404073) (88 KB) |
 |
Application Report 46 - Drugs of Abuse on Discovery HS F5 (T003046) (421 KB) |
 |
Application Report 134 - Separation of Dopamine and Related Compounds using Discovery HS F5 with Mass Spectrometric Detection (T004134) (17 KB) |
 |
Application Report 136 - Resolution of Bilirubin and Biliverdin on Discovery C18 (T004136) [Note: With MS compatible mobile phase.] (16 KB) |
 |
Application Note 179 - LC-MS CHROMASOLV® Solvents (T304179) (81 KB) |
 |
2004 Poster Presentation - Advantages of Polar, Reversed-Phase HPLC Columns for the Analysis of Drug Metabolites (T404045) (174 KB) |
 |
2004 Poster Presentation - Complementary Selectivity of Peptides on Aliphatic versus Fluorinated HPLC Phases (T404070) (158 KB) |
| TRADEMARKS |
| Discovery and CHROMASOLV are trademarks of Sigma-Aldrich Co. |