Sigma-Aldrich
Analytical / Chromatography > HPLC > Columns > Discovery® > RP Amide C16
HPLC
RP Amide C16
Discovery logo
Ordering Information
Discovery RP-Amide C16 HPLC Columns
Polar-embedded phase for unique retention and selectivity
• Excellent peak shape and efficiency
• Polar group (amide) gives less retention and different selectivity than a C18
• Not recommended for LC/MS
  Properties      
Bonded Phase:   Palmitamidopropylsilane, endcapped RP-Amide C16 structure
Silica:   Spherical, high purity (Fe <20; NA <7; Ca <7;
Ti <1; AI <1; Mg <1ppm)
Particle Size:   5µm
Pore Size:   180(Å)
Surface Area:   200m2/g
%C:   11%
Coverage:   2-3µmoles/m2
2003cat_pg24a
2003cat_pg25a
Ordering Information
5µm Discovery RP-Amide C16 HPLC Columns
ID(mm) Length(cm) Cat. No.
2.1 5 50500521
2.1 10 569320-U
2.1 12.5 569329-U
2.1 15 50501321
3.0 5 505005-30
3.0 10 569321-U
3.0 12.5 569330-U
3.0 15 505013-30
3.0 25 505064-30
4.0 5 505005-40
4.0 10 569322-U
4.0 12.5 569331-U
4.0 15 505013-40
4.0 25 505064-40
4.6 5 505005
4.6 10 569323-U
4.6 12.5 569332-U
4.6 15 505013
4.6 25 505064
10.0 25 569324-U
21.2 25 569326-U
5µm Discovery RP-Amide C16 Validation Packs*
ID(mm) Length(cm) Cat. No.
2.1 5 55705-U21
2.1 10 569802-U
2.1 15 55707-U21
4.6 5 55705-U
4.6 10 569803-U
4.6 15 55707-U
4.6 25 55709-U
* Packs include 3 columns, each from a different lof of bonded phase.
2cm Supelguard Cartridges with Discovery RP-Amide C16 Packings
  Description 2.1mm ID 3.0mm ID 4.0mm ID
  5µm Particle Size
  2 Pack
505110 59578-U 505099
  Kit triangle
505102 59577-U 505080
For 4.0mm ID or 4.6mm ID analytical columns
triangle Kit includes cartridge, stand-alone holder, tubing, 2 nuts and ferrules.
How to Contact Supelco
In USA:
Phone: 800-247-6628
  or 814-359-3441
fax: 800-325-5052
email: techservice@sial.com
Outside the USA: click here