Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > HPLC > Columns > Kromasil® > Kromasil® Cross Reference Charts
Kromasil® HPLC Products
Kromasil® Cross Reference Charts
Kromasil Eternity™ C18 Cross Reference (vs. Waters & Phenomenex)
Kromasil Polysaccharide-based Chiral Phase Cross Reference (vs. Chiral Technologies)
Kromasil Eternity™ C18 Cross Reference Chart (vs. Waters & Phenomenex) back to top
Kromasil Eternity
Catalog Number
Supelco Part
Number
Eternity   Waters Product No.*    Phenomenex Product No.** 
Particle Size (µm) I.D. (mm) Length (mm) XBridge™ XTerra® Gemini-NX Gemini™
Eternity-2.5-C18-2.1X50 K08670894 2.5 2.1 50 186003085 186000594 00B-4453-B0 00B-4439-B0
Eternity-2.5-C18-2.1X100 K08670895 2.5 2.1 100 - - - 00D-4439-B0
Eternity-2.5-C18-4.6X50 K08670896 2.5 4.6 50 186003090 186000602 00B-4453-E0 00B-4439-E0
Eternity-2.5-C18-4.6X100 K08670897 2.5 4.6 100 - - - 00D-4439-E0
Eternity-5-C18-2.1X50 K08670898 5 2.1 50 186003108 186000446 00B-4454-B0 00B-4435-B0
Eternity-5-C18-2.1X150 K08670899 5 2.1 150 186003110 186000454 00F-4454-B0 00F-4435-B0
Eternity-5-C18-4.6X50 K08670900 5 4.6 50 186003113 186000482 00B-4454-E0 00B-4435-E0
Eternity-5-C18-4.6X100 K08670901 5 4.6 100 186003115 186000486 00D-4454-E0 00D-4435-E0
Eternity-5-C18-4.6X150 K08670902 5 4.6 150 186003116 186000490 00F-4454-E0 00F-4435-E0
Eternity-5-C18-4.6X250 K08670903 5 4.6 250 186003117 186000494 00G-4454-E0 00G-4435-E0
Eternity-5-C18-10X50 K08670420 5 10 50 186002973 186001014 - 00B-4435-N0
Eternity-5-C18-10X150 K08670421 5 10 150 186002974 186001479 - 00F-4435-N0
Eternity-5-C18-10X250 K08670422 5 10 250 186003256 - - 00G-4435-N0
Eternity-5-C18-21.2X50 K08670423 5 21.2 50 186002977 186001930 00B-4454-P0-AX -
Eternity-5-C18-21.2X150 K08670424 5 21.2 150 186002979 186002379 00F-4454-P0-AX 00F-4435-P0
Eternity-5-C18-21.2X250 K08670425 5 21.2 250 186004021 - 00G-4454-P0-AX 00G-4435-P0
Eternity-5-C18-30X50 K08670426 5 30 50 186002980 186001938 00B-4454-U0-AX -
Eternity-5-C18-30X150 K08670427 5 30 150 186002984 - 00F-4454-U0-AX -
Eternity-5-C18-30X250 K08670428 5 30 250 186004025 - 00G-4454-U0-AX 00G-4435-U0
*Note: all 19mm ID columns from Waters are crossed with the 21.2mm ID Eternity columns
**Note: all 3µm Phenomenex columns are crossed with the 2.5µm Eternity columns
XBridge & XTerra are trademarks of Waters Corp.
Gemini is a trademark of Phenomenex.
Kromasil Polysaccharide-based Chiral Phase Cross Reference Chart (vs. Chiral Technologies) back to top
Kromasil
Part number
Supelco
Part No
Particle
Size (µm)
Column
I.D. x L
Chiral Technologies
Phase Part number
Kromasil phase: AmyCoat™ (Selector - amylose)
KRWP-3-AmyCoat-4.6x50 K08670346 3 4.6 mm X 50 mm Not available -
KRWP-3-AmyCoat-4.6x150 K08670344 3 4.6 mm X 150 mm Not available -
KRWP-RP-3-AmyCoat-4.6x50 (reversed-phase) K08670500 3 4.6 mm X 50 mm Not available -
KRWP-RP-3-AmyCoat-4.6x150 (reversed-phase) K08670501 3 4.6 mm X 150 mm Not available -
KRWP-5-AmyCoat-4.6x150 K08670347 5 4.6 mm X 150 mm ChiralPak™ AD-H 19324
KRWP-5-AmyCoat-4.6X250 K08670348 5 4.6 mm X 250 mm ChiralPak AD-H 19325
KRWP-5-AmyCoat-10x250 K08670605 5 10 mm X 250 mm ChiralPak AD-H 19335, 19435(SFC)
KRWP-5-AmyCoat-21.2x250 K08670606 5 20 mm X 250 mm ChiralPak AD-H 19345, 19445(SFC)
KRWP-5-AmyCoat-10x250 K08670607 5 30 mm X 250 mm ChiralPak AD-H 19375, 19475(SFC)
KRWP-10-AmyCoat-4.6x250 K08670604 10 4.6 mm X 250 mm ChiralPak AD 19025
KRWP-10-AmyCoat-10x250 K08670600 10 10 mm X 250 mm ChiralPak AD 19035
KRWP-10-AmyCoat-21.2x250 K08670601 10 20 mm X 250 mm ChiralPak AD 19045
Kromasil phase: CelluCoat™ (Selector - cellulose)
KRWP-3-CelluCoat-4.6x50 K08670372 3 4.6 mm X 50 mm Not available -
KRWP-3-CelluCoat-4.6x150 K08670370 3 4.6 mm X 150 mm Not available -
KRWP-RP-3-CelluCoat-4.6x50 (reversed-phase) K08670502 3 4.6 mm X 50 mm Not available -
KRWP-RP-3-CelluCoat-4.6x150 (reversed-phase) K08670503 3 4.6 mm X 150 mm Not available -
KRWP-5-CelluCoat-4.6x150 K08670373 5 4.6 mm X 150 mm ChiralCel™ OD-H 14324
KRWP-5-CelluCoat-4.6x250 K08670374 5 4.6 mm X 250 mm ChiralCel OD-H 14325
KRWP-5-CelluCoat-10x250 K08670614 5 10 mm X 250 mm ChiralCel OD-H 14335, 14435(SFC)
KRWP-5-CelluCoat-21.2x250 K08670615 5 20 mm X 250 mm ChiralCel OD-H 14345, 14445(SFC)
KRWP-5-CelluCoat-30x250 K08670616 5 30 mm X 250 mm ChiralCel OD-H 14375, 14475(SFC)
KRWP-10-CelluCoat-10x250 K08670609 10 10 mm X 250 mm ChiralCel OD 14035
KRWP-10-CelluCoat-21.2x250 K08670610 10 20 mm X 250 mm ChiralCel OD 14045
ChiralPak & ChiralCel are trademarks of Chiral Technologies Inc.