TSKgel® columns are available for gel filtration, gel permeation, ion exchange, reversed phase, hydrophobic interaction, hydrophilic interaction, and affinity chromatography.
TSKgel and TOYOPEARL® are widely popular columns and packing materials for the HPLC separation of proteins, peptides and other biomolecules. TSKgel columns are packed with either silica or methacrylate particles with pore sizes to accommodate a wide range of analyte molecular weights. Both particles feature surface chemistry designed to minimize sample adsorption. All popular biomolecule separation modes are covered by TSKgel columns, including size exclusion (gel filtration and gel permeation), ion exchange, affinity, reversed-phase, HIC (hydrophobic interaction) and HILIC (hydrophilic interaction) chromatography.
View the complete listing of all TSKgel columns and guard columns, and related products plus ToyoScreen® packed columns. |
|
| Technical Resources |
| Title |
| Ion Exchange Columns (45 Kb PDF) id. T494077 |
| TSKgel NPR Columns (87 Kb PDF) id. T109862 |
| TSKgel SW and SWXL Columns (56 Kb PDF) id. T494076 |
| TSKgel PW and PWXL Columns (71 Kb PDF) id. T494075 |
| TSKgel HHR Columns (68 Kb PDF) id. T496085 |
| |
|
| TSKgel SW Series Gel Filtration Columns (Silica-Based) - Ordering Information |
TSKgel SW and TSKgel SWXL columns contain silica-based, hydrophilic bonded phase packings that minimize interaction with proteins. A 30 cm TSKgel SWXL column and a 60 cm TSKgel SW column provide similar resolution, but the SWXL column requires half the analysis time. Sample capacity increases in proportion with column length.
Because TSKgel SWXL and TSKgel SW columns are silica based, they must be operated within the recommended pH range of 2.5 - 7.5. Detailed operating conditions are described in the information accompanying the columns. We recommend protecting these columns with the appropriate SWXL or SW guard column. |
| Glutathione S-Transferase |
| Column: |
TSKgel G3000SWXL, 30 cm x 7.8 mm I.D., 5 µm particles |
| Cat. No.: |
808541 |
| Mobile Phase: |
0.3M NaCl/0.05M NaH2PO4, pH 7.0 |
| Flow Rate: |
1 mL/min |
| Temp.: |
25 °C |
| Det.: |
UV, 220 nm |
| Injection: |
100 µL crude glutathione S-transferase preparation from guinea pig liver (0.7mg) |
| |
 |
|
TSKgel Column |
Particle Size (µm) |
Pore Size (Å) |
Sample MW (globular proteins) |
| Super SW2000 |
4 |
125 |
5-150 x 103 |
| G2000SWXL |
5 |
125 |
5-150 x 103 |
| G2000SW |
10 |
125 |
5-100 x 103 |
| Super SW3000 |
4 |
250 |
10-500 x 103 |
| G3000SWXL |
5 |
250 |
10-500 x 103 |
| G3000SW |
10 |
250 |
10-500 x 103 |
| G4000SWXL |
8 |
450 |
20-10,000 x 103 |
| G4000SW |
13 |
450 |
20-10,000 x 103 |
| Mobile Phase: 0.03 M NaCl in 0.1 M phosphate buffer, pH 7.0 |
|
| TSKgel PW Series Gel Filtration Columns (Polymer-Based) - Ordering Information |
TSKgel PW and TSKgel PWXL columns are used in high performance gel filtration separations of water-soluble polymers and oligosaccharides. The hydrophilic polymer matrix has excellent chemical and mechanical stability. Although commonly used with aqueous solvents, the polymer is compatible with up to 50% organic solvent.
We recommend using a TSKgel PW guard columns with G2500PW - G6000PW columns. Use a TSKgel PWXL guard column with any PWXL column.
Bulk packing can be ordered to repack PW columns and guard columns. |
TSKgel Column |
Particle Size (µm) |
Pore Size (Å) |
Sample MW (PEGS/PEOs) |
Sample MW Dextrans |
| G-Oligo-PW |
6 |
125 |
<2,000 |
- |
| G2000PW |
10 |
125 |
<2,000 |
- |
| G2500PWXL |
6 |
<200 |
<3,000 |
- |
| G2500PW |
10 |
<200 |
<3,000 |
- |
| G3000PWXL |
6 |
200 |
<50,000 |
<60,000 |
| G3000PW |
10 |
200 |
<50,000 |
<60,000 |
| G4000PWXL |
10 |
500 |
2,000-300,000 |
1,000-700,000 |
| G4000PW |
17 |
500 |
2,000-300,000 |
1,000-700,000 |
| G5000PWXL |
10 |
1,000 |
4,000-1,000,000 |
50,000-7,000,000 |
| G5000PW |
17 |
100 |
4,000-1,000,000 |
4,000-1,000,000 |
| G6000PWXL |
13 |
>1,000 |
40,000-8,000,000 |
500,000- 50,000,000 |
| G6000PW |
17 |
>1,000 |
40,000-8,000,000 |
500,000-50,000,000 |
| G-DNA-PW |
10 |
4,000 |
40,000-8,000,000 |
- |
| GMPWXL |
13 |
<100-1000 |
500-8,000,000 |
<50,000,000 |
| GMPW |
17 |
<100-1000 |
500-8,000,000 |
<50,000,000 |
Mobile Phase: Polyethylene glycols/polyethylene oxides—distilled water Mobile Phase: Dextrans—0.2 M phosphate buffer, pH 6.8 |
|
| TSKgel HHR Series Gel Permeation Columns - Ordering Information |
| TSKgel H series gel permeation columns are stable in solvents having a wide range of polarities. The particles do not swell or shrink as the solvent is changed from toluene through methanol. However, these columns cannot be used with polar solvents, such as water or water:methanol mixtures. Spherical 5 µm polystyrene/divinylbenzene particles provide a minimum of 16,000 plates per 30 cm x 7.8 mm I.D. column. Eight pore sizes are available, ranging from an exclusion limit of about 2,000 Daltons for G1000HHR columns to more than 10,000,000 Daltons for G7000HHR columns. The four mixed bed columns (H, L, M, N) feature extended linear molecular weight operating ranges for sample screening or more formal analyses. |
(see table after HXL columns for Analyte Molecular Weight Range for these columns)
|
| TSKgel HXL Series Gel Permeation Columns - Ordering Information |
| The TSKgel HHR and TSKgel HXL columns perform very similarly. The HXL columns were developed as an upgrade from the previous series, the TSKgel H. The HXL can be converted to different mobile phases for doing non-aqueous polymer separations, but it was found that the performance deteriorated when the conversion occurred. The HHR columns were developed to allow easy conversion between different mobile phases. The HHR is considered exceptionally stable, and is easily converted to other mobile phases with little loss in theoretical plates. The HXL columns are a good choice if your mobile phase of choice is THF and you do not plan on changing mobile phases. If you will change mobile phases or your mobile phase of choice is not THF, the HHR columns should be used. |
| Column |
Analyte Molecular Weight Range (Daltons) |
| G1000H |
<1,500 |
| G2000H |
<4,000 |
| G2500H |
<1.2 x 104 |
| G3000H |
<3.0 x 104 |
| G4000H |
<5.5 x 105 |
| G5000H |
<1.5 x 106 |
| G6000H |
<~1 x 107 |
| G7000H |
<~5 x 107 |
| GMH-H |
<~1 x 107 |
| GMH-L |
<1.0 x 104 |
| GMH-M |
<1.0 x 106 |
|
|
| TSKgel Affinity Columns - Ordering Information |
| High efficiency, resin-based TSKgel affinity columns (10 µm particles) separate or purify many enzymes and other proteins. The table below describes the characteristics of the columns offered. |
| |
| Characteristics of TSKgel Affinity Columns |
Immobilized Ligand |
Adsorption Capacity (per mL Gel) |
Typical Uses |
| ABA-5PW |
| p-amino benzamidine |
3-4 mg trypsin |
trypsin, bovine thrombin, urokinase, enterokinase, plasminogen activator |
| Boronate-5PW |
| m-aminophenyl boronic acid |
40 µmol sorbitol |
glycoproteins, nucleases, nucleotides, catecholamines, carbohydrates, transfer RNAs |
| Chelate-5PW |
| iminodiacetic acid |
~20 µmol Cu+2 or Zn+2 |
serum proteins, interferon, collagenase, granule protein, plasminogen activator, lactoferrin |
|
|
| TSKgel Hydrophobic Interaction Columns - Ordering Information |
Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity and allow selective binding and desorption of proteins. However, HIC operates at significantly lower binding energy and uses aqueous mobile phases. These characteristics provide a gentle technique that is less likely to disturb protein conformation. As a result, HIC generally provides better activity recovery.
TSKgel Ether-5PW, Phenyl-5PW, and Butyl-NPR resin-based columns provide a range of hydrophobicities for chromatographic optimization. Ammonium sulfate concentration can be minimized by using a very hydrophobic packing (e.g.,butyl). We recommend an Ether-5PW column for purifying very hydrophobic proteins. Ether-5PW and Phenyl- 5PW packings are based on TSKgel G5000PW resin - 10 µm particles with 1000 Å pores. Butyl-NPR packing is prepared from 2.5 µm nonporous particles, allowing rapid analyses. All three column types can be cleaned with 0.2 M NaOH. Scale-up can be performed by using corresponding Toyopearl bulk resins. |
| Proteins, mixture |
| Column: |
TSKgel Phenyl-5PW, 7.5 cm × 7.5 mm I.D., 10 µm particles |
| Cat. No.: |
807573 |
| Mobile Phase: |
0.1 M phosphate buffer, pH 7.0 plus (NH4)2SO4, 1.8 M to 0 M in 60 min |
| Flow Rate: |
1 mL/min |
| Det.: |
UV, 280 nm |
| |