Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > HPLC > Learning Center > Chromatographic Selectivity
HPLC
Chromatographic Selectivity
Chromatographic resolution is a function of column efficiency (N), retention (k) and selectivity (α). It is usually written in the form of the resolution equation:
Resolution equation
When resolution is plotted vs. these three parameters in Figure 1, it becomes apparent that selectivity has the greatest affect on resolution.
Effect of selectivity
Figure 1. Affect of Selectivity on Improving Resolution
(Figure 1 courtesy of Mao, Y. Selectivity Optimization in Liquid Chromatography Using the Thermally Tuned Tandem Column (T3C) Concept. Ph.D. Thesis, The University of Minnesota, 2001.)
Watch our instructional web seminar on HPLC Column Fundamentals to get a detailed explanation of the efficiency, retention, and selectivity terms used in the Resolution Equation. 
Additional Topics:
Developing HPLC Methods on Ascentis
Functionalized Stationary Phases for Alternate Selectivity