Sigma-Aldrich
Analytical / Chromatography > LPLC/TLC/Paper Chromatography
  • Pricing & availability is not currently available.
    Questions? Contact Technical Service
  • USA Home
  • Product Directory
  • Analytical/Chromatography
  • LPLC/TLC/Paper Chromatography
LPLC/TLC/Paper Chromatography
We offer an extensive range of products to support your low pressure column chromatography applications including an impressive selection of chromatographic media ranging from silica gels, organic adsorbents, ion-exchangers, hydrophobic interaction media and gel filtration/size exclusion media. In addition, we offer a variety of empty column housings and accessories as well as pre-packed Flash chromatography cartridges.
Our TLC product offering includes plates and foils with glass, aluminum and polyester backings in matrices ranging from silica and reversed phase silica to aluminum oxide, cellulose and polyamide. If you need TLC accessories, we also carry a selection of developing tanks, UV cabinets and hand-held UV lamps as well as TLC sprayers that make it easy for you to find everything you need all in one place.
View or download a copy of our Column Chromatography brochure
  • LPLC - Columns & Accessories - (117)
  • LPLC Separation Media - (278)
  • See also: VersaFlash® & VersaPak Products - (547)
  • Thin Layer Chromatography (TLC) - (275)
  • Chromatography Paper - (13)