Sigma-Aldrich
Analytical / Chromatography > Microbiology > Basic Ingredients > Extracts
Microbiology
Extracts
Natural based ingredients are complex ingredients and are in many cases important for the growth of fastidious microorganisms. These ingredients are mostly sources for carbohydrates, amino and fatty acids. There are also natural mixtures like bile salts which make the medium selective and inhibit the non-obligatory intestinal accompanying flora (most gram positive bacteria).
Product #  Product Name Source Add to Cart
48305 Bile Salts bile
55871 Egg powder egg
70167 Malt Extract plant
70168 Ox-bile, dehydrated, purified bile
07915 Potato Extract plant
95261 Rice extract plant
70166 Skim Milk Powder milk
Microbiology Home