Analytical / Chromatography > Microbiology > Media for Detection of E. coli O104
Microbiology Media
Media for Detection of E. coli O104
The following 3 chromogenic media used for detection of E. coli O157 also work for E coli O104, since the biochemical properties used for detection are the same in both cases. E. coli strain O104:H4 was identified as the cause of the recent EHEC problem in Germany.
Products for detection of E. coli O157 and E.coli O104
  • 72557 - HiChrome EC O157:H7 Selective Agar, Basis, BioChemika, for microbiology (Fluka)
  • 39894 - HiChrome EC O157 Agar, BioChemika, for microbiology (Fluka)
  • 80330 - HiChrome Enrichment Broth (Base) for EC O157:H7, BioChemika, for microbiology (Fluka)
e-coli
Results can be expected in 18 to 24 hours.
Note: The above products are currently
only available within the EU.
Advantages of Chromogenic Media:
  • More reliable
  • Faster results
  • Save time
  • Easy to handle
  • Save confirmation steps
  • Low cost
View all our chromogenic media in the web catalog.
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page