Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Microbiology
Kits
Staphylo Monotec Test Kit
b-Lactamase Test Kit
Staphylo Monotec Test Kit
Staphylo Monotec test kit (Fluka 08986) is a new rapid agglutination test for differentiation between Staphylococcus aureus and other Staphylococcus (see figure 1). This method has been evaluated for food material and clinical specimens. With this kit coagulase and protein A on S. aureus can be detected in one step. Due to this approach the test is highly sensitive and specific.
Staph_Kit_open_400_600
Figure 1: The rapid and reliable Staphylo Monotec test kit (easy to use)
Principle
The test reagent consists of monodisperse microparticles which are coated with IgG and fibrinogen. The reagent will detect both cell-associated coagulase and protein A on S. aureus cultured on blood agar or other recommended media for isolation of S. aureus. The negative control reagent consists of monodisperse microparticles coated with bovine serum albumin for use when the agglutination is ambiguous. On average 97% of human S. aureus isolates are coagulase positive. In addition, and independently of the coagulase property, more than 95% of all clinical S. aureus isolates have protein A in the cell wall. Because fibrinogen binds to the cell-associated enzyme coagulase, and because the Fc part of the IgG binds to protein A, a rapid agglutination occurs, when S. aureus is mixed with the Staphylo Monotec test reagent (see figure 2). USE: Bacteria and reagent are mixed for 20 seconds, and results are read immediately. Results are easy to interpret, a positive result gives clear white agglutinates against a black background (see figure 3).
Staph_principle
Figure 2: The principle of the test
StaphAggl1_400breit
Figure 3: Agglutination
Staphylo Monotec test kit has been evaluated on both human and veterinary/food material [1-2]. The presence of the control reagent is a positive feature which improves the specificity.
human
veterinary/food
Sensitivity [%] 98.6 100
Specificity [%] 99.0 98.0
Table 1: Performance of the Staphylo Monotec test kit
Click on the following product number to see further product details and to order online: 08986
References
[1] Flesland, Ø., APMIS sect. B. 95: 83-84, 1987
[2] Holme, I.J.R., Acta vet. scand. 1991, 32, 155-161
Back to top
b-Lactamase Test Kit
b-Lactams are the most widely used antibiotics, and b-lactamases are the greatest source of resistance to them. Therefore detection and identification of b-lactamase is important.
b-Lactamase Test Kit (Fluka 40561) is a rapid acidimetric test for detection of b-lactamase activity of microorganisms. The test is performed in microtitre wells. This test is based on hydrolysis of the b-lactam ring in benzylpenicillin, which results in the production of penicilloic acid. This process causes acidification of the bacterial suspension, and changes the colour of the acid-based indicator. The acidimetric test for b-lactamase activity is suitable for the detection of Haemophilus influenzae, Neisseria gonorrhoeae and Staphylococcus spp. Not suitable for the detection of the b-lactamase activity of Branhamella catarrhalis, Enteroccocus faecalis, Neisseria meningitis, Enterococcus spp. and others.
Click on the following product number to see further product details and to order online: 40561
Lactamase_testkit
Figure 4: Microtitre plate test for detecting ß-Lactamase in microorganisms
References
  1. R. Bonnet, C. Chanal, E. Ageron et al., Inducible AmpC b-Lactamase of a New Member Enterobacteriaceae, Antimicrob. Agents Chemother., 46, 3316 (2002)
  2. L. Shan, et al., Kinetic analysis of an inhibitor-resistant variant of the OHIO-1 b-lactamase, an SHV-family class A enzyme, Biochem. J., 333, 395, Printed in Great Britain (1998)
Back to top
Microbiology Home