|
Diverse biochemical reagents are known for determining certain metabolisms and to differentiate the bacteria.
Still often classical biochemical tests are used to determine microorganisms. The result can be seen by a color change. Most reactions are based on the detection of an enzyme with a certain given substrate. Also methods to detect certain metabolites by chemical reaction and or complex building techniques are used. At the end a color change gives a result which leads to more cognition of the unknown organism (see table 1).
| Cat. No. |
Detection of |
Differentiation of Organisms |
Reaction |
Color (by positive result) |
Reagent |
29333 39442 |
acetoin production
|
Klebsiella, Enterobacter |
oxidize acetoin, reacts with alpha-naphtol |
red |
Barritt's Reagent A Barritt's Reagent B |
| 07689 |
acetoin production
|
Klebsiella, Enterobacter |
oxidize acetoin, reacts with alpha-naphtol |
red |
O'Meara's Reagent |
38497 39441 |
nitrate reduction to nitrite
|
Enterobacter, E. coli, Salmonella |
reacts with sulphanilic acid, forms a azo dye with alpha-naphtylamine |
red |
Nitrate Reagent A Nitrate Reagent B |
07345
07817 |
oxidase activity
|
oxidase positives, oxidase negatives |
oxidize dimethyl-p-phenylenediamine hydrochloride, build a complex with alpha-naphtol |
dark blue |
Oxidase Reagent acc. Gaby-Hadley A Oxidase Reagent acc. Gaby-Hadley B |
| 18502 |
oxidase activity
|
oxidase positives, oxidase negatives |
oxidize N,N-dimethyl-p-phenylenediamine to Wurster's blue |
dark blue-purple |
Oxidase Reagent acc. Gordon-McLeod |
| 08714 |
strong acid production
|
E. coli, Enterobacter |
indicator reaction |
red |
Methyl Red Solution |
| 80353 |
tryptophan deaminase activity
|
Proteus |
reacts with indolepyruvic acid |
dark brown |
TDA Reagent |
| 49825 |
tryptophanase activity
|
E. coli |
reacts with indole |
blue-purple |
DMACA Reagent |
60983 67309 |
tryptophanase activity
|
E. coli |
reacts with indole |
red |
Kovac's Reagent for indoles Kovac's Reagent for indoles |
|
|
table 1: Biochemical Tests
Application example: Oxidase Reagents acc. Gaby-Hadley
These reagents can be used to perform the cytochrome c oxidase test in different methods. To differentiate organisms based on the presence of cytochrome c oxidase is one of the first and most important step by the identification of bacteria. The cytochrome c oxidase present in most gram negative bacteria oxidize dimethyl-p-phenylenediamine and alpha-naphtol a dark blue complex (indophenol blue). The test is applicable on agars in broths and on discs (see figure 1-4). Important is to do a positive and negative control with the samples.
Instructions: Grow test culture on nutrient agar slant (18-24 hours). Add 2-3 loops of reagents A and B to slant. Tilt tube to permit reagents to mix and flow over growth.
Result: Observe for color change. A positive reaction shows a dark purple blue color on growth, especially at the border of the colonies, within 2 minutes.
|
Figure 1: E. coli on a agar plate
|
Figure 2: Pseudomonas flourescence on a agar plate
|
Figure 3: paper discs impregnated with 2 drops of each reagent: left disc with E. coli; right disc with Pseudomonas flourescence
|
Figure 4: cultures with 2 drops of each reagent: left tube with E. coli; right tube with Pseudomonas fluorescence
|
Back to top Microbiology Home
|