Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
ID Membranes for Microbial Rapid Identification
New ID Membranes for Microbial
Rapid Identification (RMQ)
Membranes Functioning Like
Chromogenic and Fluorogenic Media
• Place them on established colonies for 30 seconds, then incubate for 1– 4 hours
• Outcome is similar to results on a chromogenic and/or fluorogenic medium
If you are interested in learning more and would like to see the complete range of 19 different ID membranes, please fill out the form below and you get the new ID Membranes brochure