Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Microbiology
Prepared Plates
Prepared filtration plates with a 55mm diameter, presented in a blister pack and individually protected with a separate plastic bag. Avoiding fast dehydration and contamination enhances the shelf life of the media.
prepared_plates
The use of agar plates is described in several directives related to potable water control. Such solid media allows the precise enumeration of microorganisms in water by simply incubating for a few hours. When grown on plates, the microorganisms are characterized by a typical morphology. Also, it is easy to isolate a single colony for further confirmation. These plates were specially designed to accommodate 47mm diameter water filter membranes. Save time in the preparation of your assays by using these reliable, ready to use plates.
Catalog No.
Product Name
Add to Cart
Detection of Total and Fecal Coliforms
For the detection of total coliforms incubate at 37°C, for fecal coliforms at 44°C
mFC Agar Plates
Detection of Fecal Enterococci
Enterococcus Selective Agar Plates (Slanetz-Bartley Agar; ISO 7899-2:2001)
  Total Enumeration of Aerobic Microorganisms  
Nutrient Agar Plates
Plate Count Agar Plates
Tryptic Soy Agar Plates
  Detection of Clostridia  
m-CP Agar Plates
  Detection of Pseudomonas  
Cetrimide Agar Plates
  Detection of Staphylococci  
Mannitol Salt Agar Plates
  Detection of Moulds and Yeasts  
Sabouraud 4% Glucose Agar Plates
Back to top
Microbiology Home