|
By Jvo Siegrist, Product Manager Microbiology, isiegris@europe.sial.com
HiFluoro Pseudomonas Agar Base is used as a selective medium for the isolation of Pseudomonas aeruginosa from diverse sample. Pseudomonas aeruginosa breaks the fluorogenic compound to release the fluorogen which produces a visible fluorescence under long wave UV light.
HiFluoro Pseudomonas Agar Base is formulated based on the formula described by King et al. (1) expect fluorogenic mixture. The medium is reach in essential growth nutrients. Magnesium chloride and Potassium sulfate stimulate pyocyanin production. Cetrimide (Cetyltrimethylammonium bromide) is incorporated in the medium to inhibit bacteria other than Pseudomonas aeruginosa. It acts as a quaternary ammonium compound, cationic detergent which causes nitrogen and phosphorus to be released from bacterial cells other than Pseudomonas aeruginosa. Pseudomonas aeruginosa breaks the fluorogenic compound to release the fluorogen which produces a visible fluorescence under long wave UV light.
|
 |
 |
| Pseudomonas aeruginosa on HiFluoro Pseudomonas Agar |
Pseudomonas aeruginosa on HiFluoro Pseudomonas Agar under UV light |
|
References:
- 1. E.O. King, et al., Media for the demonstration of pyocyanin and fluorescein, J. Lab. Clin. Med. 44, 301 (1954)
- G.J. Bonde, Bacterial Indicators of Water Pollution, (1962)
- L.C.D Formiga, New possibilies for the laboratory diagnosis of diphtheria, Brazilian J. Med. Biol. Res., 18, 401 (1985)
- D.J. Blazevic, M.H. Köpcke, J.M. Matsen, Incidence and identification of Pseudomonas fluorescens and Pseudomonas putida in the clinical laboratory, Appl. Microbiol., 25, 107 (1973)
For further fluorogenic media click here
Back to top Microbiology Home