Sigma-Aldrich
Analytical / Chromatography > Microbiology > Product Lines > Vegitone Media
Microbiology
Vegitone Media
No more problems with animal derived compounds in media. Approved new media with plant peptones and synthetic detergents for all kind of microorganisms.
In the Analytix Issue 3 (2005) we wrote about plant peptones as a good alternative to animal derived peptones. Meanwhile we can provide you a nice range of approved dehydrated media with plant peptones and synthetic detergent as replacement for bile salts. We can assure that this media are free from BSE and GMO! As well we have seen higher recovery rates and better growth performance with plant peptones. The new developed media are just a choice of some different kind of applications and microorganisms. If there is demand for other important formulations we are open to check the possibility to provide you a new composition. Because of less import and export problems and costs we can provide you also a lower price for this media than for the equivalent with animal derived components.
Product #  Product Name Species Add to Cart
41159 Actinomyces Agar, Vegitone
Actinomyces
40834 Actinomyces Broth, Vegitone
Actinomyces
28943 Andrade peptone water, Vegitone
Citrobacter, Escherichia coli, Klebsiella, Proteus, Providencia, Salmonella, Shigella, Yersinia & other coliforms
19057 Lactose Broth, Vegitone
Escherichia coli & Coliforms
19344 LB Agar, Vegitone
Escherichia coli, diverse other bacteria
28713 LB Broth, Vegitone
Escherichia coli, diverse other bacteria
19352 Mac Conkey Agar No 1, Vegitone
Escherichia coli, Proteus, Salmonella, Shigella & other coliforms, Staphylococci, Enterococci
38954 Malt Extract Agar modified, Vegitone
Aspergillus, Penicilliums, Saccharomyces, Pichia, Zygosaccharomyces, Candida, diverse other moulds
07012 M-FC Agar modified for Klebsiella, Vegitone
Escherichia coli, Klebsiella & other coliforms
43291 M-FC Agar, Vegitone
Escherichia coli & other coliforms
49522 M-Lauryl Sulfate Broth, Vegitone
Escherichia coli, Salmonella & other coliforms
41782 MRS Agar, Vegitone
Lactobacilli
38944 MRS Broth modified, Vegitone
Lactobacilli
04163 Nutrient Agar No 2, Vegitone
diverse moulds and bacteria
16336 Nutrient Broth No 3, Vegitone
diverse bacteria
15776 PALCAM Listeria Selective Agar, Vegitone
Listeria
59859 PALCAM Listeria Selective Enrichment Broth, Vegitone
Listeria
40893 Peptone Water, phosphate-buffered, Vegitone
Citrobacter, Escherichia coli, Klebsiella, Proteus, Providencia, Salmonella, Shigella, Yersinia & other coliforms, Staphylococci, Streptococci, Pseudomonas, diverse other bacteria
19718 Plate Count Agar, Vegitone
diverse moulds and bacteria
14432 Tryptic Soy Agar, Vegitone
diverse moulds and bacteria
41298 Tryptic Soy Broth, Vegitone
diverse moulds and bacteria
39964 Tryptone water, Vegitone
Escherichia coli, Enterococci, Klebsiella, diverse other bacteria
41960 Vegitone Infusion Broth
Coliforms, Neisseria, Staphylococci, Streptococci, diverse other bacteria
42376 Violet Red Bile Agar, Vegitone
Escherichia coli & other coliforms
53605 Violet Red Bile Glucose Agar without Lactose, Vegitone
Escherichia coli, Salmonella, Shigella & other coliforms
Back to top
References:
  1. Analytix Issue 3, Sigma-Aldrich (2005)
  2. www.sigma-aldrich.com/microbiology
Back to top
Microbiology Home