Vials
Center Drain (CD™) Vials
|
|
25% off CD™ Vials
Use Promotion Code 683
Valid Until Oct. 31, 2009 (US Customers only)
|
Special Offer – Save 25% off List Price with Promo Code 683 (offer valid to US customers only) |
The Center Drain vial has a unique conical interior bottom that allows the contents to completely drain to the center of the vial bottom. This feature permits maximum recovery of the contents, utilizing either a micropipette or syringe. The sturdy CD Vial with the patented unique feature is ideal for many micro applications. |
|
Features & Benefits:
- Enables maximum recovery of the stored sample
- Manufactured from Borosilicate Type 1 glass: safeguards against a change in the pH of the contents
- Precision: Less than 4 mL volume
- Large opening: Eliminates “bent needle” problem associated with limited volume inserts
- Maintain library samples: vials will withstand a temperature range from -70°C to 160°C
- Concentrate samples in the same vial used in an autosampler
- Popular dimension - compatible with any autosampler that uses 12 mm x 32 mm vials
|
Applications:
- Library sample storage
- Lyophilization
- Centrifugation
- Micro-mixing
- Extraction
- Concentration
- Derivatization
- Small-scale reactions
|
|
Autosampler Compatibility: Our CD vials are compatible with a wide range of autosamplers - essentially any autosampler that takes 12 mm x 32mm size vials - including the following popular brands: Agilent Series, PerkinElmer, Shimadzu, Thermo Scientific, Finnigan/Carlo Erba/Fisons, Varian, Waters, Leap Technologies
|
| Ordering Information |
| |
|
Product Description |
Pkg Qty |
List Price USD |
New Price USD |
Cat. No. |
| Replacement Caps & Septa |
| |
View Closures & Septa for CD Vials in the online catalog or email Technical Service to request additional information. |
|
| CD Vials |
| |
12 x 32 mm, 1.5 mL |
|
| |
|
Crimp top, large opening, vials only |
100 |
$81.00 |
$60.75 |
29298-U |
| |
|
Snap top, vials only |
100 |
$83.00 |
$62.25 |
29299-U |
| |
|
Snap top with pre-assembled blue cap & PTFE/septa |
100 |
$96.00 |
$72.00 |
29301-U |
| |
|
Snap top with pre-assembled bonded blue cap & PTFE/silicone septa |
100 |
$104.00 |
$78.00 |
29302-U |
| |
|
Snap top with pre-assembled bonded blue cap & pre-slit PTFE/silicone septa |
100 |
$112.00 |
$84.00 |
29303-U |
| |
9 mm vial |
|
| |
|
Clear screw top with pre-assembled bonded blue cap & PTFE/silicone septa |
100 |
$106.00 |
$79.50 |
29307-U |
| |
|
Clear screw top with pre-assembled bonded blue cap & PTFE/silicone/PTFE septa |
100 |
$118.00 |
$88.50 |
29308-U |
| |
|
Clear screw top with pre-assembled bonded blue cap & pre-slit PTFE/silicone septa |
100 |
$118.00 |
$88.50 |
29309-U |
| |
|
Amber screw top with pre-assembled bonded blue cap & PTFE/silicone septa |
100 |
$119.00 |
$89.25 |
29313-U |
| |
|
Amber screw top with pre-assembled bonded blue cap & pre-slit PTFE/silicone septa |
100 |
$133.00 |
$99.75 |
29314-U |
| |
3.8 mL, 14.75 x 45 mm, trumpet bottom |
|
| |
|
Clear vials only, screw thread |
100 |
$129.00 |
$96.75 |
29326-U |
| |
|
Clear screw top with pre-assembled bonded black cap & PTFE/silicone septa |
100 |
$159.00 |
$119.25 |
29327-U |
| |
|
Clear screw top with pre-assembled bonded black cap & pre-slit PTFE/silicone septa |
100 |
$167.00 |
$125.25 |
29328-U |
| |
|
Amber vials only, screw thread |
100 |
$136.00 |
$102.00 |
29329-U |
| |
|
Amber screw top with pre-assembled bonded black cap & PTFE/silicone septa |
100 |
$166.00 |
$124.50 |
29330-U |
| |
|
Amber screw top with pre-assembled bonded black cap & pre-slit PTFE/silicone septa |
100 |
$174.00 |
$130.50 |
29331-U |
| |
4 mL, 15 x 51 mm screw top, 13-425 thread |
|
| |
|
Kit (vial & cap & PTFE/silicone septa) |
100 |
$156.00 |
$117.00 |
29332-U |
| |
|
Vials only |
100 |
$134.00 |
$100.50 |
29334-U |
| |
5 mL, 16.75 x 51mm screw top, 15-425 thread |
|
| |
|
Kit (vial & cap & PTFE/silicone septa) |
100 |
$167.00 |
$125.25 |
29335-U |
| |
|
Vials only |
100 |
$134.00 |
$100.50 |
29336-U |
| |
10 mL, 20.75 x 61 mm screw top 18-400 thread |
|
| |
|
Kit (vial & cap & PTFE/silicone septa) |
100 |
$192.00 |
$144.00 |
29339-U |
| |
|
Vials only |
100 |
$150.00 |
$112.50 |
29340-U |
| |
20 mL, 27.75 x 67 mm screw top 24-414 thread |
|
| |
|
Kit (vial & cap & PTFE/silicone septa) |
50 |
$72.00 |
$54.00 |
29341-U |
| |
|
Vials only |
50 |
$50.00 |
$37.50 |
29342-U |
| |
40 mL, 27.75 x 103 mm screw top 24-414 thread |
|
| |
|
Kit (vial & cap & PTFE/silicone septa) |
50 |
$99.00 |
$74.25 |
29343-U |
| |
|
Vials only |
50 |
$84.00 |
$63.00 |
29344-U |
| |
60 mL, 27.75 x 143 mm screw top 24-414 thread |
|
| |
|
Kit (vial & cap & PTFE/silicone septa) |
50 |
$115.00 |
$86.25 |
29354-U |
| |
|
Vials only |
50 |
$100.00 |
$75.00 |
29355-U |
| Heavy Wall CD Vials |
| |
Clear screw top, no graduations, kit (vial & cap & PTFE/silicone septa) |
| |
|
1 mL, 13 x 41 mm, 13-425 thread |
12 |
$46.00 |
$34.50 |
29356-U |
| |
|
2 mL, 16 x 58 mm, 15-415 thread |
12 |
$60.00 |
$45.00 |
29357-U |
| |
|
2 mL, 20 x 40 mm, 20-400 thread |
12 |
$63.00 |
$47.25 |
29358-U |
| |
|
3 mL, 20 x 46 mm, 20-400 thread |
12 |
$64.00 |
$48.00 |
29359-U |
| |
|
5 mL, 20 x 61 mm, 20-400 thread |
12 |
$65.00 |
$48.75 |
29361-U |
| |
Clear screw top, with graduations, kit (vial & cap & PTFE/silicone septa) |
| |
|
1 mL, 13 x 41 mm, 13-425 thread |
12 |
$58.00 |
$43.50 |
29362-U |
| |
|
2 mL, 16 x 58 mm, 15-415 thread |
12 |
$72.00 |
$54.00 |
29363-U |
| |
|
2 mL, 20 x 40 mm, 20-400 thread |
12 |
$75.00 |
$56.25 |
29364-U |
| |
|
3 mL, 20 x 46 mm, 20-400 thread |
12 |
$76.00 |
$57.00 |
29365-U |
| |
|
5 mL, 20 x 61 mm, 20-400 thread |
12 |
$77.00 |
$57.75 |
29366-U |
| |
Amber screw top, no graduations, kit (vial & cap & PTFE/silicone septa) |
| |
|
1 mL, 13 x 41 mm, 13-425 thread |
12 |
$53.00 |
$39.75 |
29367-U |
| |
|
2 mL, 20 x 40 mm, 20-400 thread |
12 |
$75.00 |
$56.25 |
29368-U |
| |
|
3 mL, 20 x 46 mm, 20-400 thread |
12 |
$78.00 |
$58.50 |
29369-U |
| |
|
5 mL, 20 x 61 mm, 20-400 thread |
12 |
$82.00 |
$61.50 |
29370-U |
|