Analytical / Chromatography > Special Offers > 15% Discount on radiello® Air Monitoring Products
Analytical / Chromatography
15% Discount on radiello® Air Monitoring Products
Promotion Code 765
US & Canada customers only
Expires Dec 31, 2009
radiello offers many advantages over other passive (or diffusive) air monitoring devices. It has been used successfully in air sampling campaigns for current hot topics such as Indoor Vapor Intrusion, Chinese Drywall, IH/IAQ, and Ambient Air Monitoring.
Features and Benefits:
  • High, constant, reproducible uptake rate; invariable with air speed – fast and accurate sampling
  • Low detection limits with high capacity imply wide sampling range: 15 min - 30 days; 1 ppb - 1000 ppm
  • Stiff diffusive wall & water-repellent diffusive body – high tolerance, amenable to bad weather
  • Adsorbents undergo rigorous conditioning & QC – low background noise, high quality products
Special Offer - Save 15% off List Price on radiello Products and Accessories with Promo Code 765. Offer valid through Dec 31, 2009
View available radiello Products & Accessories in the web catalog.
radiello products