Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Bioanalysis: The Analysis of Compounds in Biological Matrices
Maximize the Speed & Sensitivity of Bioanalytical Assays
Sample prep and analysis of bioanalytical samples can be a challenge, whether in DMPK/ADME in pharmaceutical drug discovery, clinical diagnostics and therapeutic drug monitoring in hospitals, testing for forensics and toxicology, metabolomics, or basic biochemical research. Although each application has its unique set of analytes, matrices, and analytical objectives, the overarching need is to maximize sensitivity without compromising accuracy and laboratory throughput. We have developed unique tools to maximize speed and sensitivity throughout the entire bioanalytical workflow, especially in areas that rely on LC-MS since it has become the predominant analytical technique in bioanalysis today.
Products for LC-MS Analysis
  •  Sample Preparation: HybridSPE®-Phospholipid
    HybridSPE-PL technology is designed for the gross level removal of endogenous protein and phospholipid interferences from biological plasma and serum, prior to LC-MS or LC-MS-MS analysis.
  •  Fast HPLC, Fused-Core® Technology
    Ascentis® Express provides the high speed and high efficiency of sub-2 μm particles, but at approximately half the backpressure for the same column length.
  •  LC-MS Solvents & Reagents (CHROMASOLV®)
    Manufactured and tested to ensure the highest specifications and extremely low levels of alkali metal impurities for sensitive LC-MS. Solvent blends offer convenience and time-savings.
Supel™-Select Poymeric SPE
Supel™-Select Polymeric SPE
Hydrophilic modified styrene based polymer developed for the solid phase extraction of a broad range of compounds from aqueous samples.
Download the new Bioanalytical App for iPad
Learn how to maximize speed and sensitivity of Bioanalytical Assays with proven tools for LC-MS analysis of small molecules in biological matrices.
Download App | Download PDF
Featured Articles
TRADEMARKS: HybridSPE, Ascentis, and Supel – Sigma-Aldrich Co. LLC; CHROMASOLV – Sigma-Aldrich Laborchemikalien GmbH; Fused-Core – Advanced Materials Technology, Inc.