Sample Preparation & Purification
Pipette Tips
Pipette Tips with Zirconia (ZrO2) and Titania (TiO2) adsorbents are useful for enrichment and purification of phosphopeptides and similar bio-molecules.
C18 pipette tips offer superior binding and recovery when purifying femtomole to picomole quantities of proteins and peptides. Carbon pipette tips are used for separation, enrichment and purification of oligosaccharides and other macromolecules containing sugar moieties.
Micro-purification & Sample Enrichment
Supel-Tips contain a chromatography sorbent bed bonded at the working end of the tip with a proprietary high purity adhesive. This sorbent bed acts as a solid phase extraction medium that adsorbs molecules of interest from the sample matrix. The concentrated, desalted analytes are eluted for downstream analysis and identification with mass spectrometric and/or chromatographic techniques.
Supel-Tips offer the following separation benefits:
  • Superior recovery
  • Exceptional binding capacity and enhanced affinity
  • Excellent sorbent bed stability for cleaner samples
  • Fast and effective analyte retention/elution
supeltips pipette tips                                                          
Supel-Tips ProductsZirconia, Titania, C18, Carbon
Ordering Information
Request a FREE Pipette Tip sample pack
Technical Resources
Supel-Tips Zr Pipette Tips back to top
Product no: 54266-U
Application:
  • Sample preparation for analysis and identification of phosphopeptides
Features:
  • 10 µL micropipette Tips
  • Zirconia-silica composite sorbent
  • Sorbent: particle size = 50-60 µm; pore size = 300 Å
Supel-Tips Ti Pipette Tips back to top
Product no: 54263-U
Application:
  • Sample preparation for analysis and identification of phosphopeptides
Features:
  • 10 µL micropipette Tips
  • Titania-silica composite sorbent
  • Sorbent: particle size = 50-60 µm; pore size = 300 Å
Supel-Tips C18 Pipette Tips back to top
Product no: TPSC18
Application:
  • Sample preparation for analysis and identification of proteins, peptides
Features:
  • 10 µL micropipette Tips
  • C18 bonded spherical silica-based sorbent
  • Sorbent: particle size = 50-60 µm; pore size = 200 Å
Benefits:
  • Superior recovery of peptides and proteins
  • Exceptional binding capacity and enhanced affinity
  • Greater sorbent bed stability for cleaner samples
Supel-Tips Carbon Pipette Tips back to top
Product no: 54227-U
Application:
  • Sample preparation for analysis and identification of oligosaccharides and other macromolecules containing sugar moieties.
Features:
  • 10 µL micropipette Tips
  • Carbon adsorbent
  • Adsorbent: particle size = 50-60 µm; average pore size = 175 Å
Benefits:
  • Higher binding capacity
  • Large sample volume of 0.5-10 µL
  • Fast and effective analyte transport
Technical Resources
Publication Title Type Size (kb) No. Pages
Supel-Tips: SPE Pipette Tips for Micro-Purification/ Sample Enrichment (T407149) Brochure 697 4