Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Sample Preparation & Purification > Solid Phase Extraction > HybridSPE-PPT “In-Well” 96-well Precipitation Visual
SPE
HybridSPE-PPT “In-Well” 96-well Precipitation Visual
1. Precipitate Proteins by adding 100 µL plasma or serum to the HybridSPE-PPT plate followed by 300 µL 1% formic acid in acetonitrile. Add I.S. as necessary.
HybridSPE-PPT “In-Well” 96-well Precipitation image 1
2. Mix by vortexing/shaking HybridSPE-PPT plate or by aspirating/ dispensing with 0.5-1 mL pipette tip (e.g., TOMTEC Quadra liquid handler)
HybridSPE-PPT “In-Well” 96-well Precipitation image 2
3.
Apply vacuum.
The packed-bed filter/frit assembly acts as a depth filter for the concurrent physical removal of precipitated proteins and chemical removal phospholipids. Small molecules (e.g., pharma compounds and metabolites) pass through unretained.
HybridSPE-PPT “In-Well” 96-well Precipitation image 3
4. Resulting filtrate/eluate is free of proteins and phospholipids and ready for immediate LC-MS-MS analysis; or it can be evaporated and reconstituted as necessary prior to analysis
HybridSPE-PPT “In-Well” 96-well Precipitation Method image 4