Sigma-Aldrich
Analytical / Chromatography > Sample Preparation & Purification > Solid Phase Extraction > Method Development Kits
Solid Phase Extraction
Method Development Kits
SPE Tubes & Cartridges 
Reversed-phase SPE
Ion-exchange SPE
Normal phase SPE
Supelclean™ SPE Method Development Kits
96-Well SPE Method Development Plate
FREE SPE MultiPaks for Method Development
Application Assistance
Supelclean SPE Method Development Kits back to top
Our kits consist of an assortment of SPE phase chemistries and cartridge configurations ideal for SPE method development. The range of phase chemistries available for each kit allows the user to profile for compund retention, elution and sample matrix selectivity.
Supelclean SPE Method Development Kit Kit A Kit B Kit C Kit NP-3 Kit IX-3
Supelclean Packing Sorbent Qty. / Tube Size
LC-Si 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL
1 g/6 mL
500 mg/3 mL —
LC-8 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL — —
LC-18 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL — —
LC-CN 500 mg/3 mL 100 mg/1 mL 500 mg/6 mL — 500 mg/3 mL
LC-Diol 500 mg/3 mL 100 mg/1 mL — 500 mg/3 mL —
LC-NH2 500 mg/3 mL 100 mg/1 mL — 500 mg/3 mL 500 mg/3 mL
LC-Ph 500 mg/3 mL 100 mg/1 mL — — —
LC-SAC 500 mg/3 mL 100 mg/1 mL — — 500 mg/3 mL
LC-SCX 500 mg/3 mL 100 mg/1 mL — — 500 mg/3 mL
LC-WCX 500 mg/3 mL 100 mg/1 mL — — 500 mg/3 mL
LC-Alumina-A — — 1 g/6 mL 1 g/3 mL —
LC-Alumina-B — — 1 g/6 mL 1 g/3 mL —
LC-Alumina-N — — 1 g/6 mL 1 g/3 mL —
LC-Florisil — — 1 g/6 mL — —
Quantity each tube 6 12 3 6 12
Catalog Number 57019 57009-U 57075-U 57074-U 57073
96-Well SPE Method Development Plates back to top
96-well plateSupelco 96-well SPE Method Development Plates contain an assortment of SPE phase chemistries ideally suited for method development. The mix of phase chemistries contained within this 96-well SPE plate allows researchers to screen for analyte retention, recovery, and selectivity when achieving one’s sample prep objectives.
For ordering information, go to our On-Line Catalog
Additional Resources
Subject No.
Pages
File Size I.D.
Improve Sample Prep Selectivity through 96-well SPE Method Development Plates 3 311 Kb T206115
FREE SPE MultiPaks for Method Development back to top
SPE MultiPaks consist of an assortment of SPE phase chemistries and tube dimensions ideally suited for method development. The mix of phase chemistries available in these MultiPaks allows you to screen for optimal retention and selectivity required to achieve your sample prep objectives.
Available SPE MultiPaks
  • SupelMIP SPE MultiPaks (Molecular Imprinted Polymers)
  • Dispersive SPE (dSPE) MultiPaks
  • Supel-Select HLB SPE MultiPaks 
  • Discovery Reversed-Phase SPE MultiPaks
  • Discovery Normal-Phase SPE MultiPaks
  • Discovery Ion-Exchange SPE MultiPaks
  • Discovery DSC-MCAX (Mixed-Mode Cation Exchange) SPE MultiPak
  • Discovery DPA-6S (Polyamide) SPE MultiPak
  • Discovery Ag-Ion SPE MultiPak
  • Supelclean ENVI-Carb (Graphitized Carbon) SPE MultiPak
  • Supelclean Dual Layer SPE MultiPak (for multi-residue pesticide analysis)
  • Supelclean PSA SPE MultiPak
  • Supelclean Sulfoxide SPE MultiPak
View additional information on Discovery, Supelclean, Sample Prep Pipette Tip, and Dispersive SPE MultiPaks. You can also contact Technical Service at 814-359-3041 (800-359-3041 within the U.S.) or techservice@sial.com.
Application Assistance back to top
Refer to our library of Technical Resources for more information.
For application assistance, please use the Application Assistance Form to provide your requirements. A technical service representative will contact you to assist you in developing/troubleshooting your sample prep method.
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.