Sigma-Aldrich
Analytical / Chromatography > Sample Preparation & Purification > Solid Phase Extraction > SPE Products
  • Pricing & availability is not currently available.
    Questions? Contact Technical Service
  • USA Home
  • Product Directory
  • Analytical/Chromatography
  • Sample Preparation/Purification
  • Solid Phase Extraction (SPE)
Solid Phase Extraction (SPE)
Applications Search
Solid phase extraction (SPE) is a form of step-wise chromatography designed to extract, partition, and (or) adsorb one or more components from a liquid phase (sample) onto a stationary phase (sorbent or resin). Over the last twenty years, SPE has become the most powerful technique available for the rapid and selective sample preparation prior to analytical chromatography. SPE extends the lifetime of chromatographic systems and improves qualitative and quantitative analysis. By switching sample matrices from the original matrix to a simpler matrix environment, subsequent analysis is often simplified, and the demand placed on an analytical system is considerably lessened.
  • HybridSPE-Phospholipid - (9)
  • Supel QuE (QuEChERS) Tubes - (23)
  • SupelMIP Molecular Imprinted Polymers - (19)
  • Supel-Select SPE - (21)
  • Empore SPE disks, cartridges, and 96-well plates - (21)
  • Protein & Peptide Enrichment - (5)
  • Discovery SPE Tubes - (157)
  • Discovery SPE 96-Well Plates - (38)
  • Supelclean ENVI and Supelclean SPE Cartridges - (132)
  • SPE Method Development Plates and Kits - (6)
  • SPE Products For Combinatorial Chemistry - (26)
  • Solid Phase Extraction (SPE) Accessories - (141)
  • VersaPure Prepacked, Disposable Buchner Funnels - (32)
  • Bulk Adsorbents - (111)
  • See also: Solvents - (1477)
  • SPE Applications - (33)