Glass SPE Tubes & PTFE & SS Frits
Common in environmental analysis to reduce leachables from PP hardware and PE frits |
VersaPureTM Prepacked Büchner Funnels
 Used for large scale purification (medicinal chemistry purification) |
Reversible SPE Tubes
Reverse SPE tubes prior to elution to minimize elution volume for strongly retained compounds |