|
Solid Phase Microextraction
SPME Storage Container
|
Two devices for safely storing SPME-FFA (57592-U) or conventional SPME Fibers (57589-U) to maintain fiber conditioning or sample integrity. These also contain the spring loaded Chromline PTFE sealing mechanism. Windows in the closure of the storage container allow easy identification of the installed fiber. Please order the PTFE Sealing Cups separately in reference to the type needle of fiber intended to be stored (23 ga. bunt, 23 ga. bevel or 24 ga. blunt).
For Ordering Information click here
Technical Rescources Manual – SPME–FFA Storage Container Manual – SPME Fiber Assembly Storage Container