Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Spectroscopy
IR Solvents
Other Solvents:
IR Solvents
The increased sensitivity of modern IR-Spectroscopy demands the highest purity solvents available. Our IR Solvents have undergone rigorous purification processes in order to ensure its suitability for IR-Spectroscopy. All IR-solvents are tested for the absence of foreign band absorption in the IR-spectrum.
Please click on the product number for detailed specifications:
Catalog No.
Product Name
Purity
Pack Size
84710
Carbon disulfide for IR-spectroscopy; >99.5% (GC)
250 ml, 1 l
87030
Carbon tetrachloride for IR-spectroscopy; >99.5% (GC)
100 ml, 500 ml, 1 l
25670
Chloroform for IR-spectroscopy; >99.5% (GC),
stab. with ~0.5% ethanol
100 ml, 500 ml, 1 l
66743
Dichloromethane for IR-spectroscopy; >99.5% (GC),
stab. with cyclohexane
250 ml, 1 l
77272
Hexadecafluoroheptane pract., mixture of isomers
10 ml, 50 ml
69808
Mineral Oil for IR-spectroscopy
1 l
76235
Paraffin oil for IR-spectroscopy
100 ml, 500 ml
86970
Tetrachloroethylene for IR-spectroscopy; >99.5% (GC)
100 ml, 500 ml, 1 l
77273
Tetradecafluorohexane purum, mixture of isomers
10 ml, 50 ml
91433
1,1,2-Trichlorotrifluoroethane for IR-spectroscopy; >99.5% (GC)
250 ml, 1 l
View all IR Spectroscopy Products in the online catalog
Back to top