| |
|
Other Solvents:
|
| |
| IR Solvents |
|
The increased sensitivity of modern IR-Spectroscopy demands the highest purity solvents available. Our IR Solvents have undergone rigorous purification processes in order to ensure its suitability for IR-Spectroscopy. All IR-solvents are tested for the absence of foreign band absorption in the IR-spectrum.
Please click on the product number for detailed specifications:
|
| Catalog No. |
Product Name |
Purity |
Pack Size |
| 84710 |
Carbon disulfide |
for IR-spectroscopy; >99.5% (GC) |
250 ml, 1 l
|
| 87030 |
Carbon tetrachloride |
for IR-spectroscopy; >99.5% (GC) |
100 ml, 500 ml, 1 l
|
| 25670 |
Chloroform |
for IR-spectroscopy; >99.5% (GC), stab. with ~0.5% ethanol |
100 ml, 500 ml, 1 l
|
| 66743 |
Dichloromethane |
for IR-spectroscopy; >99.5% (GC), stab. with cyclohexane |
250 ml, 1 l
|
| 77272 |
Hexadecafluoroheptane |
pract., mixture of isomers |
10 ml, 50 ml
|
| 69808 |
Mineral Oil |
for IR-spectroscopy |
1 l
|
| 76235 |
Paraffin oil |
for IR-spectroscopy |
100 ml, 500 ml
|
| 86970 |
Tetrachloroethylene |
for IR-spectroscopy; >99.5% (GC) |
100 ml, 500 ml, 1 l
|
| 77273 |
Tetradecafluorohexane |
purum, mixture of isomers |
10 ml, 50 ml
|
| 91433 |
1,1,2-Trichlorotrifluoroethane |
for IR-spectroscopy; >99.5% (GC) |
250 ml, 1 l
|
View all IR Spectroscopy Products in the online catalog
|