Spectroscopy
NMR Solvents used in Spectroscopy
NMR Solvents / Biomolecular NMR / NMR Tubes
Spectral Viewer / Synthesis / Reference Books
Spectroscopy Home
NMR Solvents
Deuterated NMR Solvents / High Purity Solvents
NMR Reference Standards / LC-NMR Reference Standards / LC-NMR solvents
Deuterated NMR Solvents
Aldrich and Isotec have shared manufacturing methods to offer a complete line of products for the NMR industry. We continually look for ways to improve our manufacturing process so we can offer you the best NMR solvent quality possible. Take a look at these NMR solvents that now have higher isotopic enrichment.
Cat. No. Product Information Unit Sizes
Chloroform-d (previous isotopic purity 99.8 atom % D)
151823 Chloroform-d, 99.9 atom % D screw-cap 50g
    screw-cap 100g
    Sure/Seal™ 150g
    screw-cap 250g
236896 Chloroform-d, 99.9 atom % D ampule 10 x 1.0mL
441333 Chloroform-d, 99.9 atom % D ampule 10 x 0.75mL
522015 Chloroform-d, 99.9 atom % D ampule 10 x 0.60mL
308730 Chloroform-d, 99.9 atom % D ampule 5 x 0.5mL
225789 Chloroform-d, 99.9 atom % D (contains 0.03% TMS) screw-cap 100g
    Sure/Seal™ 150g
    screw-cap 500g
423661 Chloroform-d, 99.9 atom % D (contains 0.03% v/v TMS) ampule 10 x 1.0mL
151831 Chloroform-d, 99.9 atom % D (contains 1% v/v TMS) screw-cap 50g
    screw-cap 100g
    Sure/Seal™ 150g
    screw-cap 250g
Dichloromethane-d2 (previous isotopic purity 99.8 atom % D)
444324 Dichloromethane-d2, 99.9 atom % D ampule 5g
    screw-cap 10g
    screw-cap 25g
446106 Dichloromethane-d2, 99.9 atom % D ampule 10 x 0.75mL
522031 Dichloromethane-d2, 99.9 atom % D ampule 10 x 0.6mL
NEW High Purity Solvents
Aldrich now offers high purity NMR solvents that have minimum isotopic purity levels of 99.996 atom % D or better. These solvents offer the highest isotopic enrichment available for NMR applications where purity is absolutely crucial.
Cat. No. Product Information Unit Sizes
EXTRA (minimum 99.996 atom % D)
568538 Chloroform-d , EXTRA ampule 10 x 0.75mL
613398 Deuterium oxide, EXTRA Sure/Seal™ 10g
    ampule 50g
612308 (Methyl sulfoxide)-d6, EXTRA ampule 10 x 0.25mL
    ampule 10 x 0.50mL
    ampule 10 x 0.75mL
ULTRA-D (minimum 99.999 atom % D)
613401 Deuterium oxide, ULTRA-D ampule 10g
570230 Deuterium oxide, ULTRA-D ampule 10 x 0.75mL
Improved DMSO-d6 Now Lower water
We have taken steps to improve the water levels in all of our DMSO listings. Order some today and experience the difference. We have also recently added DMSO-d6, 99.9 atom % D, “Special HOH”. D2O and HOD levels are kept to a minimum to avoid deuterium exchange with easily exchangeable protons in your sample.
612324 (Methyl sulfoxide)-d6, 99.9 atom % D Special screw-cap 5g
  HOH screw-cap 25g
    screw-cap 50g
NMR Reference Standards
We offer an extensive list of NMR reference standards in a variety of tube lengths. If there is a special mix you are interested in that we do not offer, please contact our Technical Services department at 800-231-8327(USA), or your local Sigma-Aldrich sales office.
Cat. No. Product Information Tube Size
611778 1% Chloroform in acetone-d6, 99.9 atom % D 3mm x 8in
613037 1% Chloroform in acetone-d6, 99.9 atom % D 4mm x 8in
611778 1% Chloroform in acetone-d6, 99.9 atom % D 3mm x 8in
613037 1% Chloroform in acetone-d6, 99.9 atom % D 4mm x 8in
611743 1% Chloroform in acetone-d6, 99.9 atom % D 10mm x 8in
613304 2% Chloroform in chloroform-d, 99.9 atom % D 3mm x 8in
613312 2% Chlorform in chloroform-d, 99.9 atom % D 5mm x 8in
611786 5% Chloroform in acetone-d6, 99.9 atom % D 3mm x 8in
611832 5% Chloroform in acetone-d6, 99.9 atom % D 4mm Nano tube
611859 20% Chloroform in acetone-d6, 99.8 atom % D 5mm x 8in
611832 5% Chloroform in acetone-d6, 99.9 atom % D 4mm Nano tube
611859 20% Chloroform in acetone-d6, 99.8 atom % D 5mm x 8in
612510 5% 1,2-Dichlorobenzene in acetone-d6, 99.9 atom % D 5mm x 8in
612111 40% 1,4-Dioxane in benzene-d6, 99.6 atom % D 3mm x 8in
611794 40% 1,4-Dioxane in benzene-d6, 99.6 atom % D 4mm Nano tube
612073 40% 1,4-Dioxane and 5mg/mL chromium(III) acetylacetonate in benzene-d6, 99.6 atom % D 3mm x 8in
611905 40% 1,4-Dioxane and 5mg/mL chromium(III) acetylacetonate in benzene-d6, 99.6 atom % D 5mm x 8in
611824 40% 1,4-Dioxane, and 5mg/mL chromium(III) acetylacetonate in benzene-d6, 99.6 atom % D 4mm Nano tube
613045 0.1% Ethylbenzene and 0.01% TMS in chloroform-d, 99.9 atom % D 4mm x 8in
610265 0.1% Ethylbenzene and 0.01% TMS in chloroform-d, 99.9 atom % D 10mm x 8in
611808 0.1% Ethylbenzene and 0.01% TMS in chloroform-d, 99.9 atom % D 4mm Nano tube
612480 0.1% Ethylbenzene, 0.01% TMS and 0.1mg/mL chromium(III) acetylacetonate in chloroform-d, 99.9 atom % D 5mm x 8in
612936 5% Ethylbenzene and 2% TMS in chlorform-d, 99.9 atom % D 5mm x 7in
612707 45% Formamide in (methyl sulfoxide)-d6, 99.9 atom % D 3mm x 8in
611891 45% Formamide in (methyl sulfoxide)-d6, 99.9 atom % D 5mm x 8in
613339 90% Formamide in (methyl sulfoxide)-d6, 99.9 atom % D 3mm x 8in
612081 0.1mg/mL Gadolinium(III) chloride, 0.1% DSS and 1% water in deuterium oxide, 99.9 atom % D 3mm x 8in
611948 0.1mg/mL Gadolinium(III) chloride, 0.1% DSS and 1% water in deuterium oxide, 99.9 atom % D 8mm x 8in
611816 0.1mg/mL Gadolinium(III) chloride in deuterium oxide, 99.9 atom % D 4mm Nano tube
611921 25% Hexamethyldisiloxane in benzene-d6, 99.6 atom % D 5mm x 8in
613290 50% Menthol and 0.5% chromium(III) acetylacetonate in chloroform-d, 99.9 atom % D 5mm x 8in
612162 85% Phosphoric acid in deuterium oxide, 99.9 atom % D 5mm x 8in coaxial tube
611913 0.2M Potassium fluoride and 90% water in deuterium oxide, 99.9 atom % D 10mm x 8in
612685 0.05% a,a,a-Trifluorotoluene in benzene-d6, 99.6 atom % D 3mm x 8in
613185 0.05% a,a,a-Trifluorotoluene in benzene-d6, 99.6 atom % D 4mm x 8in
611179 0.0485M Triphenylphosphate in chlorform-d, 99.9 atom % D 8mm x 8in
611735 0.0485M Triphenylphosphate in chloroform-d, 99.9 atom % D 10mm x 8in
LC-NMR Reference Standards
Cat. No. Product Information Unit Size
612448 1% Chloroform in acetone-d6, 99.9 atom % D screw-cap 50mL
613029 50mM Sodium acetate and 1% water in deuterium oxide, 99.9 atom % D screw-cap 50mL
612421 1% Water in deuterium oxide, 99.9 atom % D screw-cap 250mL
LC-NMR Solvents
Cat. No. Product Information Unit Size
530891 1% Chloroform in acetone-d6, 99.9 atom % D screw-cap 50mL
613029 Acetone-d6, 99.5 atom % D GL-45 bottle 1L
530883 Acetonitrile-d3, 99.8 atom % D GL-45 bottle 1L
563102 Acetonitrile-d3, 99.8 atom % D (contains 15-20% D2O) GL-45 bottle 1L
530867 Deuterium oxide, 99.9 atom % D GL-45 bottle 1L
530875 Methyl-d3 alcohol-d, 99.8+ atom % D GL-45 bottle 1L
34955 Acetonitrile NMR CHROMASOLV® GL-45 bottle 500mL
6 x 500mL
1L
6 x 1L