NMR Solvents / Biomolecular NMR / NMR Tubes Spectral Viewer / Synthesis / Reference Books Spectroscopy Home
NMR Tubes
Aldrich NMR Tubes / Wilmad NMR Tubes / Wilmad Screw Cap NMR Tubes Norell™ NMR Sample Tubes / Shigemi Advanced NMR microtube assembly / Cross-Reference Guide
Aldrich NMR Tubes
Every Aldrich premium-quality NMR tube is inspected and gauged to meet published specifications.
- Aldrich Statement of Conformance stamped on each box of tubes
- Made of ASTM Type I borosilicate glass* for compatibility with ground joints, valves, and vacuum racks
- Precision ground and highly polished for spinning stability and highest resolution
- Thin-wall glass with round-bottoms
- Sandblasted marking spot for labelling samples
*Except disposable grade NMR tubes.
NMR TUBE SELECTION GUIDE
Choose the NMR tube grade that most closely matches the frequency (in MHz) of the spectrometer.
| Aldrich Grade |
MHz |
Applications |
Cross Ref. No. (5mm tubes) |
| Wilmad |
Kontes |
| Disposable |
30 |
Student labs, basic studies |
WG-Economy |
— |
| Series 30 |
30 |
Student labs, basic studies |
505-PS |
897200 |
| Series 200 |
60 to 270 |
Lower field studies |
506-PP |
897205 |
| Series 300 |
60 to 360 |
Routine applications |
507-PP |
897220 |
| Series 400 |
200 to 500 |
High-field, multiple-scan FT-NMR |
528-PP |
897235 |
| Series 500 |
500+ |
Ultra-high resolution |
535-PP |
897240 |
| Series 800 |
600 to 800 |
Highest resolution and performance |
541-PP |
897245 |
 |
Grade |
L (in.) |
Wall uniformity (in., TIR) |
Camber (in.) |
Cat. No. |
Add to Cart
|
| 3mm Microsample NMR tubes — 0.012in. wall thickness (Pkg=5 tubes) |
| Series 300 |
7 |
0.0020 |
0.0010 |
Z412902 |
|
| |
8 |
0.0020 |
0.0011 |
Z412910 |
|
| Series 400 |
7 |
0.0010 |
0.0005 |
Z412929 |
|
| |
8 |
0.0010 |
0.0006 |
Z412937 |
|
| Series 500 |
7 |
0.0005 |
0.00025 |
Z412945 |
|
| |
8 |
0.0005 |
0.00035 |
Z412953 |
|
| 5mm NMR tubes — 0.015in. wall thickness (Pkg=50 tubes) |
| Series 60 |
7 |
- |
- |
Z543888 |
|
| |
8 |
- |
- |
Z555908 |
|
| 5mm Thrift NMR tubes - 0.01475in. wall thickness (pkg=5 tubes) |
| Series 200 |
7 |
0.002 |
0.0005 |
Z555894 |
|
| |
8 |
0.002 |
0.0006 |
Z556092 |
|
| Series 300 |
7 |
0.0015 |
0.001 |
Z556106 |
|
| |
8 |
0.0015 |
0.001 |
Z556114 |
|
| Series 400 |
7 |
0.0005 |
0.00025 |
Z556122 |
|
| |
8 |
0.0005 |
0.00035 |
Z556130 |
|
| Series 500 |
7 |
0.0005 |
0.00025 |
Z556149 |
|
| |
8 |
0.0005 |
0.00035 |
Z556157 |
|
| 5mm Precision NMR tubes - 0.01475in. wall thickness (pkg=5 tubes) |
| Series 80 |
7 |
0.003 |
0.002 |
Z412783 |
|
| Series 200 |
7 |
0.0025 |
0.002 |
Z412791 |
|
| Series 300 |
7 |
0.002 |
0.005 |
Z412805 |
|
| |
8 |
0.002 |
0.0006 |
Z412813 |
|
| |
9 |
0.002 |
0.0007 |
Z412821 |
|
| Series 400 |
7 |
0.001 |
0.0005 |
Z412848 |
|
| |
8 |
0.001 |
0.006 |
Z412856 |
|
| Series 500 |
7 |
0.0005 |
0.00025 |
Z412864 |
|
| |
8 |
0.0005 |
0.00035 |
Z412872 |
|
| Series 800 |
7 |
0.0002 |
0.00015 |
Z412880 |
|
| |
8 |
0.0002 |
0.00015 |
Z412899 |
|
| Series 900 |
7 |
0.0002 |
0.00015 |
Z550868 |
|
| |
8 |
0.0002 |
0.00015 |
Z548243 |
|
|
|
Made of Pyrex borosilicate glass (unless otherwise noted) for compatibility with ground joints, valves, and vacuum racks. Precision ground and highly polished for spinning stability and highest resolution. Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes that are 9in. L are for making standards and for use with tip-off manifolds.
| 3mm Microsample NMR tubes - 0.012in. wall thickness (Pkg=5 tubes) |
| L(in.) |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 300MHz tubes |
| 7 |
51/25 |
307-PP-7 |
Z274321 |
|
| 8 |
51/25 |
307-PP-8 |
Z274348 |
|
| 9 |
51/25 |
307-PP-9 |
Z274356 |
|
| 360MHz tubes |
| 7 |
25/25 |
327-PP-7 |
Z271861 |
|
| 8 |
25/25 |
327-PP-8 |
Z274364 |
|
| 9 |
25/25 |
327-PP-9 |
Z274372 |
|
| 400MHz tubes |
| 7 |
25/13 |
328-PP-7 |
Z274380 |
|
| 8 |
25/13 |
328-PP-8 |
Z274399 |
|
| 9 |
25/13 |
328-PP-9 |
Z274402 |
|
| 500MHz tubes |
| 7 |
13/6 |
335-PP-7 |
Z274410 |
|
| 8 |
13/6 |
335-PP-8 |
Z274429 |
|
| 9 |
13/6 |
335-PP-9 |
Z274437 |
|
| Back to Top |
| 5mm amber NMR tubes - 0.015in. wall thickness (Pkg=1 tube) |
| L(in.) |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 100MHz tube |
| 7 |
76/51 |
505-PS-7 |
Z282251 |
|
| 360MHz tubes |
| 7 |
51/25 |
507-PP-7 |
Z282278 |
|
| 7 |
25/25 |
527-PP-7 |
Z282286 |
|
| Back to Top |
| 5mm disposable NMR tubes - 0.015in. wall thickness, N51 glass (Pkg=100 tubes) |
| L(in.) |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 100MHz tubes |
| 7 |
76/76 |
HIP-7 |
Z514144 |
|
| 8 |
76/76 |
HIP-8 |
Z514152 |
|
| Back to Top |
| 5mm economy NMR tubes - 0.015in. wall thickness. WG series made of N51 glass. (Pkg=5 tubes) |
| L(in.) |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 100MHz tubes |
| 7 |
76/76 |
WG-5-7 |
Z276278 |
|
| 8 |
76/76 |
WG-5-8 |
Z276286 |
|
| 9 |
76/76 |
WG-5-9 |
Z276294 |
|
| 7 |
76/76 |
512-5-7 |
Z276308 |
|
| 8 |
76/76 |
512-5-8 |
Z276316 |
|
| 9 |
76/76 |
512-5-9 |
Z276324 |
|
| 7 |
51/51 |
506-IM-7 |
Z514128 |
|
| 8 |
51/51 |
506-IM-8 |
Z514136 |
|
| Back to Top |
| 5mm standard NMR tubes - 0.015in. wall thickness (Pkg=5 tubes) |
| L(in.) |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 100MHz tubes |
| 7 |
76/51 |
505-PS-7 |
Z271985 |
|
| 8 |
76/51 |
505-PS-8 |
Z274445 |
|
| 9 |
76/51 |
505-PS-9 |
Z274453 |
|
| 7 |
51/51 |
506-PP-7 |
Z286028 |
|
| 8 |
51/51 |
506-PP-8 |
Z286036 |
|
| 9 |
51/51 |
506-PP-9 |
Z286044 |
|
| 200MHz tube |
| 7 |
76/51 |
503-PS-7** |
Z410357 |
|
| 360MHz tubes |
| 7 |
51/25 |
507-PP-7 |
Z271993 |
|
| 8 |
51/25 |
507-PP-8 |
Z274461 |
|
| 9 |
51/25 |
507-PP-9 |
Z272116 |
|
| 7 |
51/13 |
526-PP-7 |
Z274488 |
|
| 8 |
51/13 |
526-PP-8 |
Z274496 |
|
| 9 |
51/13 |
526-PP-9 |
Z274518 |
|
| 7 |
25/25 |
527-PP-7 |
Z274275 |
|
| 8 |
25/25 |
527-PP-8 |
Z274526 |
|
| 9 |
25/25 |
527-PP-9 |
Z274534 |
|
| 400MHz tubes |
| 7 |
25/13 |
528-PP-7 |
Z272019 |
|
| 8 |
25/13 |
528-PP-8 |
Z274291 |
|
| 9 |
25/13 |
528-PP-9 |
Z274542 |
|
| 500 MHz tubes |
| 7 |
13/6 |
535-PP-7 |
Z272027 |
|
| 7 |
13/6 |
545-PPT-7* |
Z406708 |
|
| 8 |
13/6 |
535-PP-8 |
Z274550 |
|
| 9 |
13/6 |
535-PP-9 |
Z274569 |
|
| 800 MHz tubes |
| 7 |
3.8/3.8 |
541-PP-7 |
Z412007 |
|
| 8 |
3.8/3.8 |
541-PP-8 |
Z412015 |
|
| 900 MHz tubes |
| 7 |
2.5/3.8 |
542-PP-7 |
Z412023 |
|
| 8 |
2.5/3.8 |
542-PP-8 |
Z412031 |
|
* Ultra thin wall (0.09in.) ** Medium wall (0.03in.) |
|
|
|
| 10mm NMR tubes - 0.018in. wall thickness (Pkg=5 tubes) |
| L(in.) |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 60MHz tubes |
| 7 |
NA |
513-1PS-7 |
Z274771 |
|
| 8 |
NA |
513-1PS-8 |
Z274798 |
|
| 9 |
NA |
513-1PS-9 |
Z274801 |
|
| 100MHz tubes |
| 7 |
254/51 |
Z27,468-2 |
Z272035 |
|
| 8 |
254/51 |
513-1PP-8 |
Z274690 |
|
| 9 |
254/51 |
513-1PP-9 |
Z274704 |
|
| 300MHz tubes |
| 7 |
76/38 |
513-3PP-7 |
Z272035 |
|
| 8 |
76/38 |
513-3PP-8 |
Z274712 |
|
| 9 |
76/38 |
513-3PP-9 |
Z272043 |
|
| 360MHz tubes |
| 7 |
51/25 |
513-5PP-7 |
Z274720 |
|
| 8 |
51/25 |
513-5PP-8 |
Z274739 |
|
| 9 |
51/25 |
513-5PP-9 |
Z274747 |
|
| 400MHz tubes |
| 7 |
38/13 |
513-7PP-7 |
Z272051 |
|
| 8 |
38/13 |
513-7PP-8 |
Z274755 |
|
| 9 |
38/13 |
513-7PP-9 |
Z274763 |
|
| *Tubes do not have a precision i.d. tolerance. |
|
Wilmad Screw Cap NMR Tubes
NMR tube has an open-top, screw-thread cap with a PTFE/silicone septum that permits addition of reactants or samples via syringe through the cap. Also good for sample storage. Caps and septa are replaceable. Excellent for handling air-sensitive samples and in cell perfusion studies. Tubes are 7in. L.
 |
| 5mm NMR tubes (Pkg=1 tube) |
| Product Name |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 100MHz tube |
76/51 |
505-TR-7 |
Z271918 |
|
| 100MHz tube |
51/51 |
506-TR-7 |
Z271926 |
|
| 360MHz tube |
51/25 |
507-TR-7 |
Z271934 |
|
| 400MHz tube |
25/13 |
528-TR-7 |
Z271942 |
|
| 360MHz tube |
51/25 |
507-TR-7 |
Z271934 |
|
| 400MHz tube |
25/13 |
528-TR-7 |
Z271942 |
|
| 500MHz tube |
13/60 |
535-TR-7 |
Z271950 |
|
|
8mm NMR tubes (Pkg=1 tube) |
| Product Name |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 400MHz tube |
38/13 |
513A-7TR |
Z274828 |
|
|
10mm NMR tubes (Pkg=1 tube) |
| Product Name |
Concentricity/camber (µm) |
Wilmad No. |
Cat. No. |
Add to Cart |
| 300MHz tube |
76/38 |
513-3TRA-7 |
Z271969 |
|
| 400MHz tube |
38/13 |
513-7TRA-7 |
Z271977 |
|
|
|
Replacement caps and septa for screw-cap NMR tubes
| NMR tube size (mm) |
Cat. No. |
Pkg |
Add to Cart
|
| Black open-top caps |
| 5 |
Z162531 |
144 |
|
| 10 |
Z106453 |
200 |
|
| Black solid-top caps w/white rubber liner |
| 5 |
Z188808 |
100 |
|
| 10 |
Z106410 |
200 |
|
| Black solid-top caps w/PTFE-faced rubber liner |
| 10 |
Z106437 |
200 |
|
| PTFE-faced silicone septa |
| 5 |
Z162566 |
120 |
|
| 10 |
Z106518 |
100 |
|
|
|
Norell™ NMR Sample Tubes
|
MHz
|
Length (in.)
|
Concentricity +/-(mm)
|
Camber +/- (mm)
|
Cat. No.
|
Add to Cart
|
|
3 mm Select Series™ - ASTM Type 1 Class A Glass o.d. (mm) 2.99 (+/- 0.004) i.d. (mm) 2.41 (+/- 0.006)
|
|
900
|
7
|
0.002
|
0.003
|
900
|
8
|
0.002
|
0.003
|
800
|
7
|
0.025
|
0.0038
|
800
|
8
|
0.025
|
0.0038
|
600
|
7
|
0.004
|
0.006
|
600
|
8
|
0.004
|
0.006
|
500
|
7
|
0.005
|
0.013
|
500
|
8
|
0.005
|
0.013
|
400
|
7
|
0.007
|
0.019
|
400
|
8
|
0.007
|
0.019
|
300
|
7
|
0.007
|
0.025
|
300
|
8
|
0.007
|
0.025
|
200
|
7
|
0.01
|
0.038
|
200
|
8
|
0.01
|
0.038
|
5 mm Select Series™ - ASTM Type 1 Class A Glass o.d. (mm) 4.97 (+/- 0.004) i.d. (mm) 4.20 (+/- 0.006)
|
|
900
|
7
|
0.002
|
0.003
|
900
|
8
|
0.002
|
0.003
|
800
|
7
|
0.0025
|
0.0038
|
800
|
8
|
0.0025
|
0.0038
|
600
|
7
|
0.004
|
0.006
|
600
|
8
|
0.004
|
0.006
|
500
|
7
|
0.005
|
0.013
|
500
|
8
|
0.005
|
0.013
|
400
|
7
|
0.007
|
0.019
|
400
|
8
|
0.007
|
0.019
|
300
|
7
|
0.007
|
0.025
|
300
|
8
|
0.007
|
0.025
|
200
|
7
|
0.009
|
0.035
|
200
|
8
|
0.009
|
0.035
|
5 mm Standard Series™ - ASTM Type 1 Class B, N51A Glass o.d. (mm) 4.97 (+/- 0.013) i.d. (mm) 4.20 (+/- 0.025)
|
|
600
|
7
|
0.004
|
0.006
|
600
|
8
|
0.004
|
0.006
|
500
|
7
|
0.005
|
0.013
|
500
|
8
|
0.005
|
0.013
|
400
|
7
|
0.007
|
0.019
|
400
|
8
|
0.007
|
0.019
|
300+
|
7
|
0.007
|
0.025
|
300+
|
8
|
0.007
|
0.025
|
300
|
7
|
0.01
|
0.038
|
300
|
8
|
0.01
|
0.038
|
200
|
7
|
0.01
|
0.04
|
200
|
8
|
0.01
|
0.04
|
100
|
7
|
0.02
|
0.07
|
100
|
8
|
0.02
|
0.07
|
60
|
7
|
nominal
|
nominal
|
60
|
8
|
nominal
|
nominal
|
10 mm Standard Series™ - ASTM Type 1 Class B, N51A Glass o.d. (mm) 10.00 (+/- 0.013) i.d. (mm) 8.76 (+/- 0.025)
|
|
400
|
7
|
0.005
|
0.007
|
400
|
8
|
0.005
|
0.007
|
300
|
7
|
0.02
|
0.013
|
300
|
8
|
0.02
|
0.013
|
200
|
7
|
nominal
|
nominal
|
200
|
8
|
nominal
|
nominal
|
|
|
| Back to Top |
Shigemi Advanced NMR microtube assembly
|
?Outer Sample Tube O.D. (mm)
|
Bottom L (mm)
|
Shigemi No.
|
Cat. No.
|
Add to Cart
|
|
Matched with CDCl3
|
5
|
8
|
CMS-005TB
|
5
|
12
|
CMS-005TJ
|
5
|
15
|
CMS-005TV
|
8
|
8
|
CMS-008TB
|
8
|
12
|
CMS-008TJ
|
8
|
15
|
CMS-008TV
|
Matched with CD3OD
|
5
|
8
|
MMS-005TB
|
5
|
12
|
MMS-005TJ
|
5
|
15
|
MMS-005TV
|
8
|
8
|
MMS-008TB
|
8
|
12
|
MMS-008TJ
|
8
|
15
|
MMS-005TV
|
Matched with DMSO-d6
|
5
|
8
|
DMS-005TB
|
5
|
12
|
DMS-005TJ
|
8
|
8
|
DMS-008TB
|
8
|
12
|
DMS-008TJ
|
8
|
15
|
DMS-008TV
|
5
|
15
|
DMS-005TV
|
Matched with D2O
|
5
|
8
|
BMS-005TB
|
5
|
12
|
BMS-005TJ
|
5
|
15
|
BMS-005TV
|
8
|
8
|
BMS-008TB
|
8
|
12
|
BMS-008TJ
|
8
|
15
|
BMS-008TV
|
8
|
-
|
CMS005B
|
12
|
-
|
CMS005J
|
15
|
-
|
CMS005V
|
-
|
Microtube cap |
CMSCAP
|
Matched with DMSO-d6
|
|
8
|
-
|
DMS005B
|
12
|
-
|
DMS005J
|
15
|
-
|
DMS005V
|
-
|
Microtube cap
|
DMSCAP
|
Matched with D2O
|
|
8
|
-
|
BMS005B
|
12
|
-
|
BMS005J
|
15
|
-
|
BMS005V
|
-
|
Microtube cap
|
BMSCAP
|
Cross-Reference Guide
|
?Norell™
|
Wilmad®
|
Aldrich®
|
|
Norell™
|
Wilmad®
|
Aldrich®
|
|
NORST5007
|
HIP-7
|
|
|
NORS36007
|
335-PP-7
|
|
|
NORST5008
|
HIP-8
|
|
|
NORS36008
|
335-PP-8
|
|
|
NORST5007
|
WG-5MM-ECON-7
|
|
|
NORXR557
|
505-PS-7
|
|
|
NORST5008
|
WG-5MM-ECON-8
|
|
|
NORXR558
|
505-PS-8
|
|
|
NOR5027
|
512-7
|
|
|
NORS52007
|
506-PP-7
|
|
|
NOR5028
|
512-8
|
|
|
NORS52008
|
506-PP-8
|
|
|
NOR5027
|
1206-7
|
|
|
NORS53007
|
507-PP-7
|
|
|
NOR5028
|
1206-8
|
|
|
NORS53008
|
507-PP-8
|
|
|
NOR505P7
|
1208-07
|
507-PP-9
|
|
|
NOR505P8
|
1208-08
|
|
|
NOR10017
|
513-1PP-7
|
|
|
NORXR557
|
1226-7
|
|
|
NOR10018
|
513-1PP-8
|
|
|
NORXR558
|
1226-8
|
|
|
NOR1005P7
|
513-3PP-7
|
|
|
NOR506P7
|
1228-7
|
|
|
NOR1005P8
|
513-3PP-8
|
|
|
NOR506P8
|
1228-8
|
|
|
NOR1008UP7
|
513-7PP-7
|
|
|
NOR507HP7
|
1235-7
|
|
|
NOR1008UP8
|
513-7PP-8
|
|
|
NOR507HP8
|
1235-8
|
|
|
NORS54007
|
526-PP-7
|
|
|
NOR508UP7
|
1241-7
|
|
|
NORS54008
|
526-PP-8
|
|
|
NOR508UP8
|
1241-8
|
|
|
NORS55007
|
528-PP-7
|
|
|
NOR509UP7
|
1242-7
|
|
|
NORS55008
|
528-PP-8
|
|
|
NOR509UP8
|
1242-8
|
|
|
NORS56007
|
535-PP-7
|
|
|
NORS33007
|
305-PS-7
|
|
|
NORS56008
|
535-PP-8
|
|
|
NORS33008
|
305-PS-8
|
|
|
NORS58007
|
541-PP-7
|
|
|
NORS34007
|
307-PP-7
|
|
|
NORS58008
|
541-PP-8
|
|
|
NORS34008
|
307-PP-8
|
|
|
NORS59007
|
542-PP-7
|
|
|
NORS35007
|
328-PP-7
|
|
|
NORS59008
|
542-PP-8
|
|
|
NORS35008
|
328-PP-8