Sigma-Aldrich
Analytical / Chromatography > Spectroscopy > Magnetic Resonance
Spectroscopy
Magnetic Resonance
Magnetic Resonance Imaging & Magnetic Resonance Spectroscopy
Compounds labeled with stable isotopes, such as D-Glucose-113C are used in Magnetic Resonance Imaging (MRI) techniques to render visible metabolic changes in the brain. The goal of this research is the early diagnosis of neuro-psychiatric disorders.
Researchers are also currently exploring the use of helium-3, oxygen-17, and xenon-129 gases to improve lung imaging. They are also using carbon-13 and deuterium labeled products to help elucidate metabolic pathways. Isotopic labeling provides better resolution and sensitivity than natural abundance in many imaging experiments, and the use of stable isotopes in MRI/MRS techniques is increasing. ISOTEC provides the highest enrichment and largest selection of stable isotopically labeled products for all MRI/MRS applications.
CAT NO. COMPOUND ATOM %
27,928-7 Acetic Acid-1-13C 99
27,930-7 Acetic-2-13C Acid 99
28,202-2 Acetic Acid-13C2 99
48,793-7 Algal Fatty Acid Mixture-13C 99
36,300-6 Ammonium-15N Acetate 98
  Deuterium Oxide (various grades) 99.8 to 99.999
60,629-4 Ethanol-13C2-Amine 99
60,631-6 Ethanol-2-13C-Amine 99
32,452-3 Ethyl-1-13C Alcohol 99
42,704-7 Ethyl-2-13C Alcohol 99
42,703-9 Ethyl-13C2 Alcohol 99
41,554-5 D-Galactose-1-13C 99
29,704-6 D-Glucose-1-13C 99
31,079-4 D-Glucose-2-13C 99
31,080-8 D-Glucose-6-13C 99
45,318-8 D-Glucose-1,2-13C2 99
45,319-6 D-Glucose-1,6-13C2 99
60,684-7 D-Glucose-13C6 99
55,200-3 D-Glucose-C-d7 98
60,496-8 L-Glutamic Acid-1-13C 97-99
60,512-3 L-Glutamic-2-13C Acid 99
49,001-6 L-Glutamic-3-13C Acid 99
49,292-2 L-Glutamic-5-13C Acid 99
60,501-8 L-Glutamine-1-13C 99
60,508-5 L-Glutamine-2-13C 99
60,494-1 L-Glutamine-3-13C 99
60,469-0 L-Glutamine-5-13C 99
  Helium-3 (gas) (various grades) 99.8 to 99.9999
60,147-0 Lithium-7Li Carbonate 99
60,148-9 Lithium-7Li Chloride 99.9
60,149-7 Lithium-7Li Fluoride 99
60,558-1 Octadecanoic- Acid 13C18 99
49,043-1 Oleic Acid-13C18 99
  Oxygen-17O2 (gas) (various grades) 20 to 90
29,212-5 Palmitic Acid-1-13C 99
49,275-2 Palmitic Acid-2-13C 99
48,580-2 Palmitic Acid-1,2-13C2 99
48,961-1 Palmitic Acid-1,2,3,4-13C4 99
60,557-3 Palmitic Acid-13C16 99
27,929-3 Sodium Aacetate-1-13C 99
27,931-5 Sodium Acetate-2-13C 99
28,201-4 Sodium Acetate-13C2 99
29,265-6 Sodium Butyrate-1-13C 99
48,535-7 Sodium Butyrate-2-13C 99
48,838-0 Sodium Butyrate-13C4 99
60,602-2 Sodium L-Lactate-1-13C(supplied as aqueous soln.) 99
49,004-0 Sodium L-Lactate-3-13C(supplied as aqueous soln.) 99
49,063-6 Sodium Propionate-13C3 99
49,063-6 Sodium Pyruvate-1-13C3 99
49,070-9 Sodium Pyruvate-2-13C3 99
49,072-5 Sodium Pyruvate-3-13C3 99
  Water-17O (various grades) 20 to 90
  Xenon-129 (gas) (various grades) 70 to 90
back to top
back to Spectroscopy Home