Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Peptide Analysis
MALDI Mass
MALDI-MS
MALDI (matrix-assisted laser desorption/ionization), a laser-based soft ionization method has proven to be one of the most successful ionization methods for mass spectrometric analysis and investigation of large molecules.[1,2] In addition analysis of post source decay (decay of proteins following the ionization) in many cases allows rapid sequencing of proteins. Thus MALDI has gained a crucial importance for protein analysis.
Its constituting feature is that the sample is embedded in a chemical matrix (ca. 1000 x molar excess) that greatly facilitates the production of intact gas-phase ions from large, nonvolatile, and thermally labile compounds such as proteins, oligonucleotides, synthetic polymers. and large inorganic compounds. The matrix plays a key role in this technique by absorbing the laser light energy and causing a small part of the target substrate to vaporize.
For additional information please get your free copy of our Newsletter
AnalytiX 6/2001 (Maldi, Mass Spectrometry) or download it from here
Analysis by MALDI mass spectrometry can be divided into two steps.
  • The first step involves preparing a sample by mixing the analyte with a molar excess of matrix. The typical matrix for use with ultraviolet lasers is an aromatic acid with a chromophore that strongly absorbs the laser wavelength. Other laser wavelengths are possible, in particular the mid-infrared range where the matrix can be energized by vibrational excitation; different matrix compounds must be used in this case.
  • The second step of the MALDI process involves desorption of bulk portions of the solid sample by a short pulse of laser light.
back to top
MALDI Matrices: Properties and Requirements
The MALDI matrix must meet a number of requirements simultaneously:
  • be able to embed and isolate analytes (e.g., by co-crystallization)
  • be soluble in solvents compatible with analyte
  • be vacuum stable
  • absorb the laser wavelength
  • cause co-desorption of the analyte upon laser irradiation
  • promote analyte ionization
It is believed that compounds with labile protons, such as carboxylic acids, are good MALDI matrices in the positive ion mode because they are easily able to protonate neutral analyte molecules in the plume. However, an acidic environment is not always desirable, in particular if denaturation of the tertiary structure of biomolecules should be avoided. Therefore mostly nonacidic matrices are used for protein measurements [3]. Compounds that are not easily protonated can be cationized instead, often by adding a small quantity of salt to the sample (alkali cations, and also Cu or Ag). It is also possible to detect analytes as radical cations by employing so-called electron transfer matrices [4]. More easily deprotonated compounds , such as oligonucleotides, are usually detected in negative ion mode.
The MALDI method has been developed empirically and despite its widespread use, the factors that determine success or failure of MALDI experiments are not yet fully understood. Investigations of the MALDI mechanism have recently become the focus of interest of a number of research groups [5].
back to top
Sample Preparation Techniques
In its current state, MALDI is primarily based on the laser desorption of solid matrix-analyte deposits [6]. The technique suffers from some disadvantages such as low shot-to-shot reproducibility, short sample life time and strong dependence on the sample preparation method, figure 2. A few research groups have investigated the use of liquid matrices [7], to increase sample lifetime and eliminate the search for sweet-spots, by exploiting the self-healing properties of the sampling position through molecular diffusion. Table 1 shows different sample preparation methods.
Table 1: Different sample preparation methods
Solid Matrix
Liquid Matrix
Special preparations
Dried-droplet
Chemical liquid
Solid supports
Vacuum-drying
Particle-doped (two-phase) liquid
MALDI on 2D-gels
Crushed-crystal
Chemical-doped liquid
Insoluble samples
Fast-evaporation
-
-
Overlayer
.
-
Sandwich
-
-
Spin-coating
.
-
Slow-crystallization
-
-
Electrospray
.
-
Quick and dirty
-
-
Matrix precoated targets
.
-
back to top
Applications and Choice of Matrix
The most important applications of MALDI mass spectrometry are (in decreasing order of importance): peptides and proteins, synthetic polymers, oligonucleotides, oligosaccharides, lipids, inorganics. Numerous matrices have been found for these and other classes of compounds; a summary is given in figure 1. Although electrospray ionization (ESI) is somewhat competitive and certainly complementary, MALDI remains the method of choice in several key areas, particularly proteomics. ESI mass spectra include many peaks of multiply charged ions which can complicate the interpretation of the spectra of complex samples. Also, the ion current is distributed over a range of m/z values, sometimes compromising sensitivity. The sensitivity of ESI is also severely reduced by the presence of salts, impurities, and organic buffers which are more easily tolerated by MALDI.
Table 2. Guide to Sample Preparation with FLUKA products and Sigma Calibration Kits
Catalog No.
Product Name
Abbreviation
Determined Molecule
Peptide, Protein
70990
alpha-Cyano-4-hydroxycinnamic acid
CHCA
Mass < 10 kDa
alpha-Cyano-4-hydroxycinnamic acid Diethylamine salt  
ioniq matrix
alpha-Cyano-4-hydroxycinnamic acid Butylamine salt  
ioniq matrix
85429
Sinapic acid
SA
Mass > 10 kDa
54793
2-(4-Hydroxyphenylazo) benzoic acid
HABA
-
76154
2-Mercapto-benzothiazole
-
-
14078
Succinic acid
-
IR-Laser
37468
2,6-Dihydroxy acetophenone
-
UV-Laser
46278
Ferulic acid
-
UV-Laser
60018
Caffeic acid
-
UV-Laser
49771
Glyerol
-
Liquid matrix
72681
4-Nitroaniline
-
Liquid matrix
Universal MALDI-matrix
-
Mixed matrix
03565
MALDI Validation Set polystyrene
-
-
03596
MALDI Validation Set poly(butyl acrylate)
-
-
03597
MALDI Validation Set PS, PMMA, PDMS, PEG, PSS
-
-
03598
MALDI Validation Set polyethylene glycol
-
-
03599
MALDI Validation Set poly(methyl methacrylate)
-
-
Oligonucleotide
91928
2,4,6-Trihydroxy acetophenone
THAP
Mass < 3.5 kDa
56197
3-Hydroxy picolinic acid
HPA
Mass > 3.5 kDa
10678
Anthranilic acid
-
-
72311
Nicotinic acid
-
-
84228
Salicylamide
-
-
Synthetic polymer
57288
Trans- 3-indoleacrylic acid
IAA
Non-polar
10608
Dithranol
DIT
-
85707
2,5-Dihydroxy benzoic acid
DHB
Polar
14078
Succinic acid
-
IR-Laser
Organic molecules
85707
2,5-Dihydroxy benzoic acid
DHB
-
Universal MALDI-matrix
-
Mixed matrix
59927
Isovanillin
-
-
Carbohydrates
85707
2,5-Dihydroxybenzoic acid
DHB
-
70990
alpha-Cyano- 4-hydroxy cinnamic acid
CHCA
-
07336
3-Aminoquinoline
DHB
-
91928
2,4,6-Trihydroxyacetophenone
THAP
Acidic
10608
Dithranol
DIT
Lipids
Dendrimers
85429
Sinapic acid
SA
-
10608
Dithranol
THAP
DIT
85429
Sinapic acid
SA
Fullerenes
87884
T-2-(3-(4-t- Butyl-phenyl)- 2-methyl- 2-propenylidene) malononitrile
DCTB
Inorganic molecules
55433
1-Isoquinolinone
-
Oligosaccharide
Peptide, Proteins
MSCAL1
ProteoMassTMPeptide&Protein Calibration Kit
-
Mass range of standards: 757 Da to 66,430 Da
Peptide
MSCAL2
ProteoMassTM Peptide Calibration Kit
-
Mass range of standards: 757 Da to 3,494 Da
Proteins
MSCAL3
ProteoMassTM Peptide&Protein Calibration Kit
-
Mass range of standards: 5,730 Da to 66,430 Da
Back to Top
Table 3. List of Products
Catalog No.
Product Name
Application
Package 1
Package 2
07336
3-Aminoquinoline carbohydrates
1 g
5 g
10678
Anthranilic acid oligonucleotide
1 g
-
06788
Bencoic acid polymer
10x10 mg
60018
Caffeic acid peptide, proteins
1 g
5 g
06773
Caffeic acid peptide, proteins
10x10mg
70990
alpha-Cyano-4-hydroxycinnamic acid peptide, proteins
250 mg
1 g
55272
alpha-Cyano-4-hydroxycinnamic acid peptide, proteins
10x25mg
39468
alpha-Cyano-4-hydroxycinnamic acid (ultra pure) peptide, proteins
10x10mg
10608
Dithranol synthetic polymer, lipids
1 g
5 g
54238
Dithranol synthetic polymer, lipids
10x10mg
37468
2,6-Dihydroxyacetophenone peptide, proteins
1 g
5 g
85707
2,5-Dihydroxybenzoic acid synthetic polymers / carbohydrates
250 mg
1 g
39318
2,5-Dihydroxybenzoic acid (ultra pure) synthetic polymers / carbohydrates
10x10mg
46278
Ferulic acid peptide, proteins
1 g
5 g
18077
Ferulic acid peptide, proteins
10x10 mg
49771
Glycerol peptide, proteins
1 ml
5 ml
54793
2-(4-Hydroxyphenylazo)benzoic acid peptide, proteins
1 g
5 g
56197
3-Hydroxypicolinic acid oligonucleotide
250 mg
1 g
55433
1-Isoquinolinol oligosaccharide
1 g
5 g
59927
Isovanillin organic molecules
1 g
5 g
76154
2-Mercapto-benzothiazole peptide, proteins
250 mg
1 g
72311
Nicotinic acid oligonucleotide
250 mg
1 g
38372
Nicotinic acid oligonucleotide
10X10 mg
72681
4-Nitroaniline peptide, proteins
250 mg
1 g
39497
4-Nitroaniline peptide, proteins
10x10 mg
84228
Salicylamide oligonucleotide
1 g
5 g
06789
Salicylamide oligonucleotide
10x10 mg
85429
Sinapic acid peptide, proteins
dendrimers / fullerenes
1 g
5 g
78867
Sinapic acid dendrimers / fullerenes
10x10mg
49508
Sinapic acid (ultra pure) dendrimers / fullerenes
10x10 mg
14078
Succinic acid synthetic polymer
1 g
5 g
66912
Succinic acid synthetic polymer
10x10 mg
87884
T-2-(3-(4-t-Butyl-phenyl)-2-methyl-2- propenylidene)malonitrile inorganic molecules
1 g
5 g
57288
Trans-3-indoleacrylic acid synthetic polymer
1 g
5 g
91928
2,4,6-Trihydroxyacetophenone carbohydrates / oligonucleotides
1 g
5 g
41711
2,4,6-Trihydroxyacetophenone carbohydrates / oligonucleotides
10x10 mg
03599
MALDI Validation Set poly(methyl methacrylate) peptide, proteins
1 set
-
03598
MALDI Validation Set polyethylene glycol peptide, proteins
1 set
-
03565
MALDI Validation Set polystyrene peptide, proteins
1 set
-
03597
MALDI Validation Set PS, PMMA, PDMS, PEG peptide, proteins
1 set
-
MSCAL1 (Sigma)
ProteoMassTM Peptide&Protein Calibration Kit
Peptide, proteins
Kit
-
MSCAL2 (Sigma)
ProteoMassTM Peptide Calibration Kit Peptide, proteins
Kit
-
MSCAL3 (Sigma)
ProteoMassTM Peptide&Protein Calibration Kit Peptide, proteins
Kit
-
Literature
  1. Karas, M.; Bachmann, D.; Bahr, U.; Hillenkamp, F. Int. J. Mass Spectrom. Ion Proc. 1987, 78, 53-68.
  2. Tanaka, K.; Waki, H.; Ido, Y.; Akita, S.; Yoshida, Y.; Yoshida, T. Rapid Comm. Mass Spectrom. 1988, 2, 151 - 153..
  3. Fitzgerald, M. C.; Parr, G. R.; Smith, L. M. Anal. Chem. 1993, 65, 3204 - 3211.
  4. McCarley, T. D.; McCarley, R. L.; Limbach, P. A. Anal. Chem. 1998, 70, 4376-4379.
  5. Dale, M.J.; Knochenmuss, R.; Zenobi R. Rapid Commun. Mass Spectrom. 1997, 11, 136-142.
  6. Chapman, J. R. Methods in Mol. Biol.; Humana Press: Totowa, NJ, 1996; Vol. 61.
  7. Zenobi, R.; Knochenmuss, R. Mass Spectrom. Rev. 1999, 17, 337 – 366.
Back to Top