|
Mass Spectroscopy
MS Markers
|
|
|
Mass Spectrometry Standards
Some highly fluorinated organic compounds are useful as reference materials for mass spectrometry. The reason for their utility in MS applications is three-fold:
- perfluorinated compounds are generally very stable
- they have high volatility compared to non-fluorinated materials ot the same molecular mass
- there is little spectral overlap between the fluorinated reference and non-fluorinated sample, making the interpretation easier
MS Standards
| Cat. No. |
Products |
Description |
CAS No. |
Package Size |
| 77298 |
Perfluorotetracosane |
p.a. standard for mass spectroscopy |
1766-41-2 |
500 mg |
| 77299 |
Heptacosafluorotributylamine |
p.a. standard for mass spectroscopy |
311-89-7 |
5, 25 ml |
| 93332 |
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine |
purum p.a. standard for mass spectroscopy; >98.0% (GC) |
915-76-4 |
100, 500 mg |
| 93383 |
2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine |
purum p.a. standard for mass spectroscopy; >98.0% (NMR) |
858-46-8 |
100, 500 mg |
| 93384 |
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine |
purum p.a. for mass spectroscopy; >97.0% (GC) |
21674-38-4 |
100, 500 mg |
| 86311 |
Bis(pentafluorophenyl)-phenyl-phosphine |
p.a. standard for mass spectroscopy |
5074-71-5 |
100, 500 mg |
| 78744 |
Hexadecafluoro-2-butyltetra-hydrofuran |
p.a. standard for mass spectroscopy |
335-36-4 |
1, 5 g |
| 96187 |
Fluorinated -phosphazenes |
p.a. standard for mass spectroscopy |
|
250 mg |
| 03565 |
MALDI Validation Set Polystyrene |
Mp 500-70'000, certified according to DIN |
|
1 each |
| 03596 |
MALDI Validation Set Poly(butyl acrylate) |
Mp 700-80'000, certified according to DIN |
|
1 each |
| 03597 |
MALDI Validation Set PS, PMMA, PDMS, PEG and PSS |
Mp 5'000-20'000 certified according to DIN |
|
1 each |
| 03598 |
MALDI Validation Set Polyethylene glycol |
Mp 500-20'000 certified according to DIN |
|
1 each |
| 03599 |
MALDI Validation Set Poly(methyl methacrylate) |
Mp 500-70'000 certified according to DIN |
|
1 each |
Back to top
Back to Mass Spectroscopy home
Back to Analytical Standards home
|
| |
| |
| |
| |
|