Sigma-Aldrich
Analytical / Chromatography > Supelco - Analytical > Equity® Products
Supelco
Equity® Products
eQuity Logo eQuity Products Image
Equity-1
Equity-5
Equity-1701
The performance you demand... the service you deserve... from the Company you trust.
Supelco's Equity Capillary GC Columns deliver the capillary GC column performance you demand for your nonpolar applications.
Significant improvements in the polymer chemistry are at the heart of the enhanced performance you will receive with our new Equity Capillary GC Columns. The polymer improvements result in better bonding, higher thermal stability, and superior product reproducibility.
The Resolution You Need
For accurate identification, reliable quantitation, and confidence in your results choose Equity Capillary GC Columns. The consistently high resolution you demand and the service you deserve.
The Analyte Response You Require
For lower detection limits and less instrument downtime you can depend on Equity Capillary GC Columns. The reproducible analyte response you demand and the service you deserve.
The Low Bleed You Expect
For great sensitivity, reliable identification, and increased sample throughput rely on Equity Capillary GC Columns. The consistent low bleed performance you demand and the service you deserve.
The Column Life You Count On
To increase your productivity and reduce your instrument downtime use Equity Capillary GC Columns. The durable, consistent column life you demand and the service you deserve.
Ordering Information
Equity-1 Capillary GC Columns
phase:   bonded; poly(dimethylsiloxane)
temp. limits:  
0.25 and 0.32mm ID:   -60°C to 325/350°C
0.53mm ID:   -60°C to 300/320°C (£1.5µm Df)
    -60°C to 260/280°C (>1.5µm Df)
This column meets USP G1, G2, and G9 requirements.
I.D. Length (m) Df (µm) b Cat. No.
0.10mm        
  15 0.10 250 28039-U
0.20mm        
  12 0.33 152 28041-U
  25 0.33 152 28042-U
  10 1.2 42 28043-U
0.25mm        
  30 0.10 625 28044-U
  15 0.25 250 28045-U
  30 0.25 250 28046-U
  60 0.25 250 28047-U
  15 1.0 63 28048-U
  30 1.0 63 28049-U
  60 1.0 63 28050-U
  100 1.0 63 28052-U
0.32mm        
  20 0.10 800 28053-U
  15 0.25 320 28054-U
  30 0.25 320 28055-U
  60 0.25 320 28056-U
  30 1.0 80 28057-U
  60 1.0 80 28058-U
  100 1.0 80 28060-U
  30 2.0 40 28061-U
  30 5.0 16 28062-U
  60 5.0 16 28063-U
0.53mm        
  15 0.10 1325 28064-U
  30 0.10 1325 28065-U
  15 0.5 265 28067-U
  30 0.5 265 28068-U
  15 1.0 132 28069-U
  30 1.0 132 28071-U
  15 1.5 88 28072-U
  30 1.5 88 28073-U
  60 1.5 88 28074-U
  15 3.0 44 28075-U
  30 3.0 44 28076-U
  60 3.0 44 28077-U
  15 5.0 27 28079-U
  30 5.0 27 28081-U
  60 5.0 27 28082-U
back to top
Equity-5 Capillary GC Columns
phase:   bonded; poly(5% diphenyl/95% dimethylsiloxane)
temp. limits:  
0.25 and 0.32mm ID:   -60°C to 325/350°C
0.53mm ID:   -60°C to 300/320°C (£1.5µm Df)
    -60°C to 260/280°C (>1.5µm Df)
This column meets USP G27,and G36 requirements.
I.D. Length (m) Df (µm) b Cat. No.
0.10mm        
  15 0.10 250 28083-U
0.20mm        
  15 0.20 250 28084-U
  30 0.20 250 28085-U
  60 0.20 250 28086-U
  12 0.33 152 28087-U
0.25mm        
  15 0.25 250 28088-U
  30 0.25 250 28089-U
  60 0.25 250 28090-U
  30 0.5 125 28092-U
  15 1.0 63 28093-U
  30 1.0 63 28094-U
  60 1.0 63 28095-U
0.32mm        
  15 0.25 320 28096-U
  30 0.25 320 28097-U
  60 0.25 320 28098-U
  30 0.32 250 28099-U
  30 0.5 160 28195-U
  10 1.0 80 28337-U
  30 1.0 80 28199-U
  60 1.0 80 28251-U
0.53mm        
  15 0.5 265 28252-U
  30 0.5 265 28259-U
  60 0.5 265 28263-U
  30 1.0 133 28264-U
  15 1.5 88 28265-U
  30 1.5 88 28267-U
  15 3.0 44 28268-U
  30 3.0 44 28269-U
  15 5.0 27 28278-U
  30 5.0 27 28279-U
  60 5.0 27 28293-U
back to top
Equity-1701 Capillary GC Columns
phase:   bonded; poly(14% cyanopropylphenyl/86% dimethylsiloxane)
temp. limits:  
0.25 and 0.32mm ID:   subambient to 280°C
0.53mm ID:   subambient to 260°C
This column meets USP G46 requirements.
I.D. Length (m) Df (µm) b Cat. No.
0.10mm        
  15 0.10 250 28343-U
0.25mm        
  15 0.25 250 28371-U
  30 0.25 250 28372-U
  60 0.25 250 28373-U
  15 1.0 63 28374-U
  30 1.0 63 28378-U
  60 1.0 63 28379-U
0.32mm        
  15 0.25 320 28381-U
  30 0.25 320 28382-U
  60 0.25 320 28384-U
  15 1.0 80 28386-U
  30 1.0 80 28387-U
  60 1.0 80 28388-U
0.53mm        
  15 0.5 265 28389-U
  30 0.5 265 28391-U
  15 1.0 132 28393-U
  30 1.0 132 28394-U
  15 1.5 88 28395-U
  30 1.5 88 28396-U
 back to top
Equity Brochure
The Equity Capillary GC Column brochure includes applications that are relevant to today’s analysts working within the industrial, clinical, environmental, food and beverage, and pharmaceutical markets. We present applications on our improved Equity capillary GC columns including pesticides, solvents, fuel oils, phenols, drugs of abuse, and blood alcohol analyses. Equity columns offer the consistent resolution, analyte response, low bleed, and column life you demand for your applications. To receive a copy by mail, please contact Supelco Technical Service at 800-359-3041/814-359-3041. To download the 52 page brochure (1,256 Kb PDF), click here.