Syringes
SGE
SGE syringes are available for GC and LC manual injection, and for the major brands of GC and HPLC autosamplers.
Use of microliter volume syringes often presents two concerns - delicate plungers, and proportionally large sample hold-up in the needle. SGE syringe options featuring Plunger Protection, Guided Plunger and SuperfleX® Plungers help prevent bending or kinking of the plunger. Syringes with the Plunger-in-Needle feature permit the injection of very small sample volume.
We can also supply other SGE syringes not presented below.
Microvolume Liquid Syringes
  Standard Plunger
  Extended barrel (Guided Plunger)
  Plunger Protection
  Plunger-in-Needle
  SuperfleX® Flexible Plunger
Gas Tight Syringes
  Autosampler Syringes
New! icon eVol® hand-held automated analytical syringes from SGE
SGE Syringe Tips & Frequently Asked Questions (FAQ)
Microvolume Liquid Syringes
Standard Plunger back to top
p001077
Fixed needle, removable needle, bevel tip
Syringe Volumes: 25 µL, 50 µL, 100 µL, 250 µL, 500 µL
  • Stainless steel plunger is individually fitted to the barrel
  • Plunger is not replaceable
Extended Barrel (Guided Plunger) Syringes back to top
SGE guided plunger syringe
Fixed needle, removable needle, bevel tip
Syringe Volumes: 5 µL, 10 µL
  • Extended barrel guides the plunger during injection
  • Robust design: suitable for technician or student use, multiple users
Plunger Protection Syringes back to top
sge plunger protection syringe
Fixed needle, removable needle, bevel tip
Syringe Volumes: 5 µL, 10 µL
  • Extended protection tube from top of the flange helps prevent plunger bending during injection.
Plunger-in-Needle Syringes back to top
plunger in needle syringe
Plunger in needle, cone and bevel tips
Syringe Volumes: 0.5 µL, 1 µL, 5 µL
  • Plunger extends to needle tip; sample is contained only in the needle
  • Ideal for dispensing very small liquid volumes
SuperfleX Flexible Plunger Syringes back to top
sge superflex syringe
Fixed needle, removable needle, bevel and cone tips
Syringe Volumes: 5 µL, 10 µL
  • Indestructible plunger; made from Titanium/Nickel
  • Ideal for student use
Gas Tight Syringes
  • Constructed from borosilicate glass, with PTFE plunger tips
  • Dispensed Volume accuracy of ± 1%
Syringe Volume: less than 1 mL back to top
Fixed needle, removable needle, both with bevel tip
Syringe volumes: 10 µL, 25 µL, 50 µL, 100 µL, 250 µL, 500 µL
Syringe Volume: 1 mL - 100 mL back to top
sge luer lock gas tight syringe
Removable needle, bevel tip
Syringe volumes: 1 mL, 2.5 mL, 5 mL, 10 mL
Fixed luer lock
Syringe volumes: 1 mL, 2.5 mL
Removable luer lock
Syringe volumes: 5 mL, 10 mL, 25 mL, 50 mL, 100 mL
SuperfleX is a registered trademark of SGE International