|
Titration
HYDRANAL® Karl Fischer Reagents
|
|
Our premium quality Riedel-de Haën products for pyridine-free water determination by Karl Fischer titration (KF titration) comprise one- and two-component reagents for volumetric and reagents for coulometric Karl Fischer titrations. HYDRANAL E-type reagents are ethanol-based. Also available are water standards for titre standardization, pH buffering reagents, special reagents for aldehydes and ketones and other solubilizing agents.
 Products:
 Special Offers:
 Tool Box:
|
|
 Learning Center:
|
 |
|
|
| |
|
| |
|
|
HYDRANAL® is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH Riedel-de Haën® is a registered trademark of Honeywell International, Inc., licensed to Sigma-Aldrich |