Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
HYDRANAL®
Additional Reagents
For many applications pure methanol like HYDRANAL®-Methanol dry is used to dissolve the samples. It is however often necessary to alter the working medium to suit the individual sample. Non-polar samples require chloroform or a long-chained alcohol as solubilizer whereas polar samples and many foodstuffs necessitate the addition of formamide. Acidic and basic samples often must be neutralised in order to reach the ideal pH-value or to suppress side-reactions.
Sigma-Aldrich developed several working media which can be used for most of the usual applications and samples. Additionally we several solubilizers and buffers. The product range is completed by HYDRANAL®-Humidity Absorber. This drying agent is suitable to reduce air moisture which is one of the most common sources of error in the Karl Fischer titration.
View Product List and Ordering Information
Literature
HYDRANAL®-Buffer Base- Lab Info in English
HYDRANAL®-Buffer Base - Lab Info in German
Back to top
Back to HYDRANAL®-home