Sigma-Aldrich
Analytical / Chromatography > Titration > HYDRANAL® Karl Fischer Reagents > Coulometric Reagents
HYDRANAL®
Coulometric Reagents
Ordering Information
Composition
Coulometry normally necessitates two reagent solutions, an anolyte and a catholyte. The anolyte is added to the anodic compartment and the catholyte to the cathodic compartment.
HYDRANAL®-Coulomat A reagents are used as anolyte. Every anolyte contains iodide and a sulphur dioxide/imidazole buffer in a suitable solvent. HYDRANAL®-Coulomat C reagents are used as catholytes.
Toxicity
HYDRANAL®-Coulomat G-Types have been developed as anolyte as well as catholyte.
These reagents are:
  • Pyridine free.
  • Chloroform free.
  • Carbon tetrachloride free.
  • 2-methoxyethanol free.
HYDRANAL®-Coulomat G reagents contain methanol as the only hazardous component. HYDRANAL®-Coulomat E is based on ethanol.
Stability
All reagents have a shelf life of at least five years, as long as the bottles remain unopened.
Advantages of the coulometric titration
  • Easy to use.
  • High accuracy.
  • Suitable for the determination of 10 µg - 10 mg of water.
  • Reagents available with and without halogenated hydrocarbons.
HYRDRANAL videos
Learn more - watch our
HYDRANAL videos
View Product List and Ordering Information
Literature
English
Karl Fischer titration The Coulometric Cell
HYDRANAL®-Coulomat Oil A new reagent for the coulometric water determination in oils
Deutsch
Karl-Fischer-Titration Die Coulometriezelle
HYDRANAL®-Coulomat Oil Ein neues Reagenz für die coulometrische Wasserbestimmung in Ölen
Back to top
Back to HYDRANAL®-home