Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Karl Fischer Titration Reagents (HYDRANAL®)
Analytical Reagents Search
HYDRANAL reagents for water determination by Karl Fischer titration fulfill all requirements of practical laboratory analysis, and enable analysts to choose the best reagent for their specific application and instrumentation.
The product range consists of one-component and two-component reagents for volumetric determinations, coulometric reagents, and specially designed reagents for the determination of water in aldehydes, ketones and other difficult-to-solubilize substances. The product range is completed by water standards for titer determination (water equivalent) and buffer solutions for maintaining an ideal pH range between 5 and 7.
Browse through our HYDRANAL video selection and the basics of KF titration to review different variations of Karl Fischer titration procedures.