Sigma-Aldrich
Analytical / Chromatography > Titration > HYDRANAL® Karl Fischer Reagents > HYDRANAL®-E Types
HYDRANAL®
HYDRANAL®-E Types
HYDRANAL is going green ... Karl Fischer titration with ethanol-based reagents
At the center of the continuous developments is the replacement of toxic methanol which is widely used as solvent in the titration
vessel or as solvent for Karl Fischer reagents by the non-toxic ethanol. HYDRANAL-E types represent the first ethanol-based Karl Fischer reagent line for the volumetric and the coulometric titration.
Advantages of HYDRANAL-E types
  • Compared with methanol based Karl Fischer reagents handling is less dangerous for users.
  • Reagents meet requirements of ISO EN DIN 14001.
  • Improved solubility of long-chained hydrocarbons.
  • Ketones like acetone can be titrated in HYDRANAL-CompoSolver E without side-reaction.
  • Compared with the traditional two-component reagents the ethanol-based reagents have the same titration speed, identical shelf life and titre parameters.
  • HYDRANAL-Coulomat E can be used as one-component-reagent for coulometric cells with diaphragm.
Order a laboratory report
Please state the report number from the table below. For any questions, please contact the HYDRANAL®-Technical Helpline.
View Product List and Ordering Information
Back to top
Back to HYDRANAL®-home